Merck
  • Toward large scale roll-to-roll production of fully printed perovskite solar cells.
Toward large scale roll-to-roll production of fully printed perovskite solar cells.
Advanced materials (Deerfield Beach, Fla.) (2015-01-13)
Kyeongil Hwang, Yen-Sook Jung, Youn-Jung Heo, Fiona H Scholes, Scott E Watkins, Jegadesan Subbiah, David J Jones, Dong-Yu Kim, Doojin Vak
ABSTRACT
Fully printed perovskite solar cells are demonstrated with slot-die coating, a scalable printing method. A sequential slot-die coating process is developed to produce efficient perovskite solar cells and to be used in a large-scale roll-to-roll printing process. All layers excluding the electrodes are printed and devices demonstrate up to 11.96% power conversion efficiency. It is also demonstrated that the new process can be used in roll-to-roll production.
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%