Merck
  • Endocrinization of FGF1 produces a neomorphic and potent insulin sensitizer.
Endocrinization of FGF1 produces a neomorphic and potent insulin sensitizer.
Nature (2014-07-22)
Jae Myoung Suh, Johan W Jonker, Maryam Ahmadian, Regina Goetz, Denise Lackey, Olivia Osborn, Zhifeng Huang, Weilin Liu, Eiji Yoshihara, Theo H van Dijk, Rick Havinga, Weiwei Fan, Yun-Qiang Yin, Ruth T Yu, Christopher Liddle, Annette R Atkins, Jerrold M Olefsky, Moosa Mohammadi, Michael Downes, Ronald M Evans
ABSTRACT
Fibroblast growth factor 1 (FGF1) is an autocrine/paracrine regulator whose binding to heparan sulphate proteoglycans effectively precludes its circulation. Although FGF1 is known as a mitogenic factor, FGF1 knockout mice develop insulin resistance when stressed by a high-fat diet, suggesting a potential role in nutrient homeostasis. Here we show that parenteral delivery of a single dose of recombinant FGF1 (rFGF1) results in potent, insulin-dependent lowering of glucose levels in diabetic mice that is dose-dependent but does not lead to hypoglycaemia. Chronic pharmacological treatment with rFGF1 increases insulin-dependent glucose uptake in skeletal muscle and suppresses the hepatic production of glucose to achieve whole-body insulin sensitization. The sustained glucose lowering and insulin sensitization attributed to rFGF1 are not accompanied by the side effects of weight gain, liver steatosis and bone loss associated with current insulin-sensitizing therapies. We also show that the glucose-lowering activity of FGF1 can be dissociated from its mitogenic activity and is mediated predominantly via FGF receptor 1 signalling. Thus we have uncovered an unexpected, neomorphic insulin-sensitizing action for exogenous non-mitogenic human FGF1 with therapeutic potential for the treatment of insulin resistance and type 2 diabetes.
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
NIH 3T3 Cell Line murine, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 93061524, mouse embryo, Fibroblast
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
Heparin sodium porcine mucosa
293 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85120602, human kidney (embryonic), Epithelial
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture