Skip to Content
Merck
All Photos(1)

Documents

00538

Supelco

Chlorophyll b

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H70MgN4O6
CAS Number:
Molecular Weight:
907.47
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

InChI

1S/C55H71N4O6.Mg/c1-12-38-35(8)42-27-43-36(9)40(23-24-48(61)65-26-25-34(7)22-16-21-33(6)20-15-19-32(5)18-14-17-31(3)4)52(58-43)50-51(55(63)64-11)54(62)49-37(10)44(59-53(49)50)28-46-39(13-2)41(30-60)47(57-46)29-45(38)56-42;/h12,25,27-33,36,40,51H,1,13-24,26H2,2-11H3,(H-,56,57,58,59,60,62);/q-1;+2/p-1/b34-25+;/t32?,33?,36-,40-,51?;/m0./s1

InChI key

NSMUHPMZFPKNMZ-FCFLHPMBSA-M

General description

Chlorophyll b belongs to the class of porphyrins conjugated with magnesium. It absorbs light energy mostly in the blue region and along with chlorophyll a, it is responsible for the green color of plants and green algae.

Application

This analytical standard can also be used as follows:

  • Fourier transform infrared (FTIR) spectroscopy-based quantification of chlorophylls (a & b) and pheophytins (a & b) from five brands of green tea samples
  • Development of a high-performance liquid chromatography-diode array detection-mass spectrometric (HPLC-DAD-MS) method for the separation and determination of chlorophylls along with their derivatives in Taraxacum formosanum samples
  • Analysis of 14 dietary supplement samples for the quantification of chlorophylls, its degradation products, carotenoids, and pheophytins by a UV-vis spectrophotometric method following their separation by high-performance thin-layer chromatography (HPTLC)
  • Evaluation of the effect of pressure, temperature, CO2 flow rate, and a polar co-solvent on the supercritical fluid extraction (SFE) of carotenoids and chlorophylls from microalgae Scenedesmus obliquus for use in food processing
  • Simultaneous measurement of chlorophylls (a & b) and carotenoids (β-carotene and lycopene) in fruit samples of strawberry, apricot, and raspberry by two UV-vis spectrophotometric methods

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara I Faria et al.
Polymers, 12(3) (2020-03-18)
Understanding the conditions affecting cyanobacterial biofilm development is crucial to develop new antibiofouling strategies and decrease the economic and environmental impact of biofilms in marine settings. In this study, we investigated the relative importance of shear forces and surface hydrophobicity
Ming-Hua Liang et al.
Journal of agricultural and food chemistry, 65(15), 3190-3197 (2017-04-04)
Dunaliella tertiolecta, a halotolerant alga, can accumulate large amounts of neutral lipid, which makes it a potential biodiesel feedstock. In this study, neutral lipids of D. tertiolecta induced by different salinities or N or P starvation were analyzed by thin-layer chromatography
Yuting Sheng et al.
BioMed research international, 2020, 2854795-2854795 (2020-07-01)
As one of the most important micronutrients, iron (Fe) plays a critical role in various metabolic processes during plant growth and development. However, the molecular mechanisms towards Fe metabolism and nutrition in fruit trees are largely unknown. In this study
Sagheer Ahmad et al.
BMC plant biology, 20(1), 145-145 (2020-04-09)
Axillary bud is an important agronomic and economic trait in cut chrysanthemum. Bud outgrowth is an intricate process controlled by complex molecular regulatory networks, physio-chemical integrators and environmental stimuli. Temperature is one of the key regulators of bud's fate. However
Erika C Pierattini et al.
Tree physiology, 38(1), 109-118 (2017-12-12)
Surfactants are widely used detergent ingredients and, thanks to their chemical properties, they are applied for remediation of sites polluted by heavy metals and organic contaminants, both in soil flushing and in phytoremediation. However, their direct effects on tree physiology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service