Skip to Content
Merck
All Photos(1)

Key Documents

MAB3890

Sigma-Aldrich

Anti-PPAR α Antibody

ascites fluid, Chemicon®

Synonym(s):

PPAR alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PPARA(5465)

General description

Peroxisome proliferator-activated receptors (PPARs) are nuclear hormone receptors that can be activated by a variety of compounds including fibratus, thiazolidinediones, prostaglandins and fatty acids. Three PPAR subtypes, designated PPARα, PPARβ (also designated PPARδ) and PPARγ, have been described. PPARs promote transcription by forming heterodimers with members of the retinoid X receptor (RXR) family of steroid receptors and binding to specific DNA motifs termed PPAR-response elements (PPREs). PPARα is abundant in primary hepatocytes where it regulates the expression of proteins involved in fatty acid metabolism. PPARβ is the most widely distributed subtype and is often expressed at high levels. PPARγ is predominantly seen in adipose tissue where it plays a critical role in regulating adipocyte differentiation.

Specificity

Reacts with Peroxisome Proliferator Activated Receptor alpha (PPARalpha). No cross reactivity with PPARbeta or PPARgamma.

Immunogen

Synthetic peptide from amino acids 18-34 of mouse PPARalpha.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-PPAR α Antibody (Mouse Monoclonal Antibody) validated in ELISA, WB, ICC to detect PPAR alpha also known as Peroxisome Proliferator Activated Receptor α.
Western blot: 1:500-1:5,000

Immunocytochemistry: 1:500-1:5,000

ELISA: 1:500-1:5,000

Optimal working dilutions must be determined by end user.

Physical form

Ascites fluid. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
3T3 whole cell lysate

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Demin Cai et al.
Journal of cellular physiology, 236(6), 4387-4402 (2020-11-14)
Nonalcoholic-fatty-liver-disease (NAFLD) is the result of imbalances in hepatic lipid partitioning and is linked to dietary factors. We demonstrate that conjugated linoleic acid (CLA) when given to mice as a dietary supplement, induced an enlarged liver, hepatic steatosis, and increased
Hao-Yu Liu et al.
Frontiers in nutrition, 8, 711398-711398 (2021-11-02)
Scope: Disruptions of circadian rhythm cause metabolic disorders and are closely related to dietary factors. In this study, we investigated the interplays between the dietary conjugated linoleic acid (CLA)-induced hepatic steatosis and the circadian clock regulation, in association with lipid
Anthony R Soltis et al.
Scientific reports, 7(1), 174-174 (2017-03-12)
Diet plays a crucial role in shaping human health and disease. Diets promoting obesity and insulin resistance can lead to severe metabolic diseases, while calorie-restricted (CR) diets can improve health and extend lifespan. In this work, we fed mice either
Hao-Yu Liu et al.
Liver international : official journal of the International Association for the Study of the Liver, 42(6), 1449-1466 (2022-02-21)
Disruption of lipid metabolism is largely linked to metabolic disorders, such as hypercholesterolemia (HCL) and liver steatosis. While cholesterol metabolic re-programmers can serve as targets for relevant interventions. Here we explored the dietary conjugated linoleic acids (CLA)-induced HCL in mice
Ning Jia et al.
Nutrition & metabolism, 15, 86-86 (2018-12-18)
Non-alcoholic fatty liver disease (NAFLD) is a chronic and progressive liver disease with an increased risk of morbidity and mortality. However, so far no specific pharmacotherapy has been approved. Gynostemma pentaphylla (Thunb.) Makino (GP) is a traditional Chinese medicine that is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service