Skip to Content
Merck
All Photos(1)

Documents

CP18

Sigma-Aldrich

Anti-Collagen Type II (Ab-1) Mouse mAb (II-4C11)

liquid, clone II-4C11, Calbiochem®

Synonym(s):

Collagen Type II antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

II-4C11, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human, bovine, rabbit

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL2A1(1280)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with P3-X-63-Ag8-LUI mouse myeloma cells. Recognizes the ~130 kDa type II collagen protein.
Recognizes the ~130 kDa type II collagen protein. May react with other collagens. When used at high concentrations higher bands corresponding to aggregates may also be detected.
This Anti-Collagen Type II (Ab-1) Mouse mAb (II-4C11) is validated for use in Frozen Sections for the detection of Collagen Type II (Ab-1).

Immunogen

Human
purified collagen type II

Application

Frozen Sections (1-10 µg/ml)

Immunoblotting (1-5 µg/ml, see application references)

Paraffin Sections (1-10 µg/ml, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM sodium phosphate buffer, 0.1% BSA, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Kumagai, J. et al. 1994 J. Anat.185, 278.
Kivrikko, K,I. 1993 Ann. Med.25, 113.
Sandell, L.J., et al. 1991 J. Cell. Biol.114, 1307.
Ryan, M.C., and Sandell, L.J. 1990 J. Biol. Chem.265, 10334.
Mayne, R., and Burgeson, R.E. 1987 Structure and Function of Collagen Types,Academic Press, Orlando, FL
May react with other collagens. When used at high concentrations higher bands corresponding to aggregates may also be detected. This antibody may also react with other classes of collagen at a concentration higher than 1 µg/ml. In immunoblotting, a major band of ~130 kDa is obtained. Antibody should be titrated for optimal results in individual systems.

Legal Information

Manufactured by Daiichi Fine Chemical Co., Ltd. Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng-Zheng Zhang et al.
Science translational medicine, 11(487) (2019-04-12)
Reconstruction of the anisotropic structure and proper function of the knee meniscus remains an important challenge to overcome, because the complexity of the zonal tissue organization in the meniscus has important roles in load bearing and shock absorption. Current tissue
Chang Qing et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 44(4), 303-310 (2011-04-14)
Chondrocytes and bone marrow mesenchymal stem cells (BMSCs) are frequently used as seed cells in cartilage tissue engineering. In the present study, we determined if the co-culture of rabbit articular chondrocytes and BMSCs in vitro promotes the expression of cartilaginous
Johannes Kupka et al.
International journal of molecular sciences, 21(6) (2020-03-22)
Healthy and degenerating intervertebral discs (IVDs) are innervated by sympathetic nerves, however, adrenoceptor (AR) expression and functionality have never been investigated systematically. Therefore, AR gene expression was analyzed in both tissue and isolated cells from degenerated human IVDs. Furthermore, human
Shiping Shi et al.
Experimental and therapeutic medicine, 12(5), 3161-3166 (2016-11-25)
Osteoarthritis (OA) is a joint disease, and few treatments to date have been able to delay OA progression. The degradation of collagen type II (COL2) in the cartilage matrix is an important initiating factor for OA progression; the upregulation of
Han-Jun Kim et al.
Oxidative medicine and cellular longevity, 2020, 5915481-5915481 (2020-10-09)
Intervertebral disc degeneration (IVDD) is a common cause of lower back pain. Programmed cell death (PCD) including apoptosis and autophagy is known to play key mechanistic roles in the development of IVDD. We hypothesized that the nucleus pulposus cells that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service