Skip to Content
Merck
All Photos(3)

Documents

SAB4500366

Sigma-Aldrich

Anti-Collagen II antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CO2A1, chondrocalcin, collagen, collagen α-1 (II), type II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 141 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL2A1(1280)

General description

Anti-Collagen II Antibody detects endogenous levels of total Collagen II protein.
Collagen type II α 1 chain (COL2A1) gene is mapped to human chromosome 12q13.11–q13.12. COL2A1 belongs to collagen superfamily. The encoded protein is characterized with three identical α chains forming homotrimers. Collagen II is the major component of cartilage.

Immunogen

The antiserum was produced against synthesized peptide derived from human Collagen II.

Immunogen Range: 101-150

Application

Anti-Collagen II, N-Terminal antibody produced in rabbit has been used for:
  • Western blotting
  • Immunofluorescence
  • Immunohistological analyses
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (2 papers)

Biochem/physiol Actions

Mutation of collagen type II α 1 chain (COL2A1), leads to a variety of clinical manifestations such as skeletal dysplasia, short stature, and sensorial defects, collectively known as type II collagenopathies. Function of COL2A1 in articular cartilage is to form highly organized fibers, which are essential for the mechanical properties of the tissue.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouna Barat-Houari et al.
Human mutation, 37(1), 7-15 (2015-10-08)
Mutations in the COL2A1 gene cause a spectrum of rare autosomal-dominant conditions characterized by skeletal dysplasia, short stature, and sensorial defects. An early diagnosis is critical to providing relevant patient care and follow-up, and genetic counseling to affected families. There
E V Isaeva et al.
Sovremennye tekhnologii v meditsine, 15(2), 5-16 (2023-06-30)
The aim of the study was to compare type I collagen-based and methacryloyl gelatin-based (GelMA) hydrogels by their ability to form hyaline cartilage in animals after subcutaneous implantation of scaffolds. Chondrocytes were isolated from the costal cartilage of newborn rats
Elena V Isaeva et al.
International journal of molecular sciences, 23(5) (2022-03-11)
The aim of this study was to verify the applicability of high-concentration collagen-based bioink with MSC (ADSC) and decellularized ECM granules for the formation of cartilage tissue de novo after subcutaneous implantation of the scaffolds in rats. The printability of
Guokun Chen et al.
Frontiers in pharmacology, 11, 527744-527744 (2020-10-13)
The purpose of this study was to elucidate the role of the circadian gene Bmal1 in human cartilage and its crosstalk with the MAPK/ERK signaling pathway in temporomandibular joint osteoarthritis (TMJ-OA). We verified the periodical variation of the circadian gene
Shiping Shi et al.
Experimental and therapeutic medicine, 24(2), 526-526 (2022-07-16)
Osteoarthritis (OA) is a chronic condition caused by cartilage degradation, and there are currently no effective methods for preventing the progression of this disease; gene therapy is a relatively novel method for treating arthritis. Decreased collagen type II (Col2) expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service