Skip to Content
Merck
All Photos(1)

Key Documents

AP124

Sigma-Aldrich

Goat Anti-Mouse IgG Antibody

2 mg/mL, Chemicon®

Synonym(s):

Anti-Mouse IgG, HRP-Conjugated Goat Anti-Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with Mouse IgG gamma chain, as well as the light chains from all mouse immunoglobulin classes.Monospecific on IEP against whole mouse serum.

Application

Goat anti-Mouse IgG Antibody detects level of Mouse IgG & has been published & validated for use in ELISA, IP & WB.

Quality

Single band on IEP at a concentration of 10 mg/mL when run against rabbit anti goat whole serum.

Physical form

Liquid in 0.02M Phosphate Buffer, 0.25M Sodium Chloride and 0.1% Sodium Azide. Filtered through a 0.45 micron membrane filter.

Storage and Stability

Store at +2-8°C in undiluted aliquots for up to six months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinghao Wang et al.
International journal of medical sciences, 18(1), 18-28 (2021-01-05)
Cardiac hypertrophy (CH) is a major risk factor for heart failure accompanied by maladaptive cardiac remodeling. The role and potential mechanism of neuropeptide Y (NPY) in CH are still unclear. We will explore the role and the mechanism of NPY
Chongwen Wang et al.
Sensors and actuators. B, Chemical, 345, 130372-130372 (2021-07-06)
Rapid and accurate diagnosis of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) and influenza A virus (FluA) antigens in the early stages of virus infection is the key to control the epidemic spread. Here, we developed a two-channel fluorescent immunochromatographic
Kathleen M Lennon et al.
Journal of extracellular vesicles, 8(1), 1685634-1685634 (2019-11-20)
Biofluid-accessible extracellular vesicles (EVs) may represent a new means to improve the sensitivity and specificity of detecting disease. However, current methods to isolate EVs encounter challenges when they are used to select specific populations. Moreover, it has been difficult to
S Seo et al.
Neuroscience, 192, 28-36 (2011-07-23)
Agmatine, the decarboxylated metabolite of l-arginine, is considered to be a novel putative neurotransmitter. Recent studies have demonstrated that endogenous agmatine may directly participate in the processes of spatial learning and memory. Agmatine-immunoreactivity has been observed within synaptic terminals of
Neda Rafat et al.
Biosensors, 10(10) (2020-10-22)
Electrochemical immunosensors (EIs) integrate biorecognition molecules (e.g., antibodies) with redox enzymes (e.g., horseradish peroxidase) to combine the advantages of immunoassays (high sensitivity and selectivity) with those of electrochemical biosensors (quantitative electrical signal). However, the complex network of mass-transfer, catalysis, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service