Skip to Content
Merck
All Photos(1)

Documents

DC05L

Sigma-Aldrich

Goat Anti-Mouse IgG Alkaline Phosphatase Conjugate

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoaffinity purified goat polyclonal antibody adsorbed against human, bovine, and horse serum proteins and conjugated to alkaline phosphatase. Recognizes mouse IgG.
This Goat Anti-Mouse IgG Alkaline Phosphatase Conjugate is validated for use in ELISA Immunoblotting for the detection of Mouse IgG.

Immunogen

Mouse

Application

ELISA (1:5000-1:100,000)

Immunoblotting (1:5000-1:100,000, colorimetric; 1:10,000-1:200,000, chemiluminescence)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 250 mM NaCl, 10 mM Tris-HCl, 15 mg/ml BSA, pH 8.0.

Reconstitution

Reconstitute the antibody with 1 ml H₂O. Dissolve by gently mixing at 4°C, since extensive mixing will denature antibodies. Following reconstitution, aliquot and freeze (-70°C). Avoid freeze/thaw cycles of solutions. Alternatively, glycerol (to a final concentration of 50%) may be added and the reconstituted antibody then stored at -20°C. Please note, under these conditions the concentration of the antibody and the buffer salts will be one-half of the original concentration.

Other Notes

To ensure the quality of these reagents each lot is tested with several of our primary antibodies. Antibody specificity is tested by immunoelectrophoresis and shown to be specific for mouse IgG and light chains common to most mouse immunoglobulins. No cross-reactivity with non-immunoglobulin serum proteins has been detected. The antibody has also been tested by ELISA and/or adsorbed to ensure minimal cross-reactivity with human, bovine, and horse serum proteins, but the antibody may exhibit cross-reactivity with immunoglobulins from other species. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Su et al.
Plant direct, 6(5), e403-e403 (2022-06-07)
In Arabidopsis, phytochrome (phy) A, phyB, and cryptochrome 1 (cry1) are representative far-red, red, and blue light photoreceptors, respectively. Members of the SUPPRESSOR OF PHYA-105 (SPA) protein family (SPA1-SPA4) form E3 ubiquitin ligase complexes with CONSTITUTIVE PHOTOMORPHOGENIC1 (COP1), which mediates
Subrata K Pore et al.
Journal of cellular biochemistry, 121(1), 804-815 (2019-08-14)
Anticancer drugs exert their effects on cancer cells by deregulating many pathways linked to cell cycle, apoptosis, etc. but cancer cells gradually become resistive against anticancer drugs, thereby necessitating the development of newer generation anticancer molecules. N-end rule pathway has
Sujan Kumar Mondal et al.
Molecular pharmaceutics, 13(7), 2507-2523 (2016-05-18)
Many cancers including the late stage ones become drug-resistant and undergo epithelial-to-mesenchymal transition (EMT). These lead to enhanced invasion, migration, and metastasis toward manifesting its aggressiveness and malignancy. One of the key hallmarks of cancer is its overdependence on glycolysis
Pritha Agarwalla et al.
Molecular therapy oncolytics, 3, 16020-16020 (2016-08-25)
Recent study has shown that N-end rule pathway, an ubiquitin dependent proteolytic system, counteracts cell death by degrading many antisurvival protein fragments like BCLxL, BRCA1, RIPK1, etc. Inhibition of the N-end rule pathway can lead to metabolic stabilization of proapoptotic
Eliedonna Cacao et al.
Biointerphases, 8(1), 9-9 (2014-04-08)
We have developed a technique for the high-resolution, self-aligning, and high-throughput patterning of antibody binding functionality on surfaces by selectively changing the reactivity of protein-coated surfaces in specific regions of a workpiece with a beam of energetic helium particles. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service