Skip to Content
Merck
All Photos(1)

Key Documents

AP187

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, Species Adsorbed

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Goat anti-Rabbit IgG Antibody, Species Adsorbed is an antibody against Rabbit IgG for use in ELISA, IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Lundby et al.
Scandinavian journal of medicine & science in sports, 27(12), 1627-1637 (2017-02-07)
Interindividual variation in running and cycling exercise economy (EE) remains unexplained although studied for more than a century. This study is the first to comprehensively evaluate the importance of biochemical, structural, physiological, anthropometric, and biomechanical influences on running and cycling
Win-Ping Deng et al.
The spine journal : official journal of the North American Spine Society, 14(10), 2488-2499 (2014-05-06)
Recent advanced studies have demonstrated that cytokines and extracellular matrix (ECM) could trigger various types of neural differentiation. However, the efficacy of differentiation and in vivo transplantation has not yet thoroughly been investigated. To highlight the current understanding of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service