Skip to Content
Merck
All Photos(1)

Key Documents

8.20528

Sigma-Aldrich

Diphenylamine

for synthesis

Synonym(s):

Diphenylamine, N-Phenylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-539-4
NACRES:
NA.22

vapor pressure

0.0003 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

630 °C (DIN 51794)

potency

1120 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

302 °C/1013 hPa

mp

53-54 °C

transition temp

flash point 153 °C (DIN 51758)

solubility

0.05 g/L

density

1.16 g/cm3 at 20 °C

bulk density

610 kg/m3

storage temp.

no temp limit

SMILES string

N(c2ccccc2)c1ccccc1

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Application

  • Design of hole transport materials: New small derivatives of 2,2′-bithiophene, possibly involving Diphenylamine as a building block, were rationally designed to improve the performance of perovskite solar cells, demonstrating Diphenylamine′s role in advancing photovoltaic technology (Adadi et al., 2024).
  • High-efficiency hyperfluorescent OLEDs: Diphenylamine may be implicated in the development of hybridized local and charge transfer dendrimers, aimed at achieving near-unity exciton utilization in solution-processed OLEDs, highlighting its potential in high-performance electronic displays (Yin et al., 2024).
  • Chemical detection technologies: A benzoxazole-triphenylamine conjugated fluorogenic probe was developed for the specific detection of sarin gas mimic diethylchlorophosphate, illustrating Diphenylamine′s utility in sensitive and specific chemical sensors (Sultana et al., 2024).
  • Electrochemiluminescence emitters: Diphenylamine-based emitters were tailored for efficient electrochemiluminescence, employing tripropylamine as a co-reactant, contributing to the field of electrochemical sensors and displays (Morgan et al., 2023).
  • Near-infrared fluorophores development: Research on chlorinated rylenecarboximide fluorophores, potentially involving Diphenylamine, focused on efficient near-infrared applications, which are critical for advanced imaging and diagnostic techniques (Wu et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 52 °C
Melting range (upper value): ≤ 55 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service