Skip to Content
Merck

242586

Diphenylamine

ACS reagent grade, ≥99%

Synonym(s):

DPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
204-539-4
MDL number:
Beilstein/REAXYS Number:
508755
Assay:
≥99%
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Diphenylamine, ACS reagent, ≥99%

grade

ACS reagent

Quality Segment

vapor density

5.82 (vs air)

vapor pressure

1 mmHg ( 108 °C)

assay

≥99%

autoignition temp.

1175 °F

ign. residue

≤0.03%

bp

302 °C (lit.)

mp

50-53 °C (lit.)

solubility

alcohol: passes test

anion traces

nitrate (NO3-): passes test

suitability

passes test for sensitivity to nitrate

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Application

Diphenylamine may be used in the preparation of diphenylamine reagent, which is employed for the quantitative estimation of nucleic acid (DNA) from various biological sources by colorimetric method.
Diphenylamine can undergo palladium-catalyzed intramolecular cyclization to form carbazole. It may also be used as a starting material in the synthesis of triphenylamine.


Still not finding the right product?

Explore all of our products under Diphenylamine


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

target_organs

Kidney,Liver,spleen

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library