Skip to Content
Merck
All Photos(1)

Key Documents

8.01756

Sigma-Aldrich

Benzaldehyde

for synthesis

Synonym(s):

Benzaldehyde, Bitter almond oil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O
CAS Number:
Molecular Weight:
106.12
MDL number:
UNSPSC Code:
12352114
EC Index Number:
202-860-4
NACRES:
NA.22
Assay:
≥99.0% (GC)
bp:
179 °C/1013 hPa
vapor pressure:
1.3 hPa ( 20 °C)

vapor pressure

1.3 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

190 °C

potency

1300 mg/kg LD50, oral (Rat)
>1250 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-8.5 % (v/v)

pH

5.9 (1 g/L in H2O)

bp

179 °C/1013 hPa

mp

-26 °C

transition temp

flash point 64 °C

solubility

3.3 g/L

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Related Categories

Application

Benzaldehyde can be used as a reactant to synthesize:
  • β-amino ketone derivatives by Cu nanoparticles catalyzed one-pot, three-component Mannich reaction with various aromatic ketones and amines.
  • Enantioselective 1-phenyl-1-propanol by asymmetric addition of diethylzinc in the presence of fenchone-based derivatives as catalysts.
  • Benzoylborane by reacting with a boryl Grignard reagent (borylmagnesium).
  • Diethyl benzylidenemalonate by piperidine catalyzed reaction with diethyl malonate.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.044 - 1.046
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions under microwave conditions
Tetrahedron Organic Chemistry Series (2005)
Novel one-pot Cu-nanoparticles-catalyzed Mannich reaction
Kidwai M, et al.
Tetrahedron Letters, 50(12), 1355-1358 (2009)
Synthesis, structure of borylmagnesium, and its reaction with benzaldehyde to form benzoylborane.
Makoto Yamashita et al.
Journal of the American Chemical Society, 129(31), 9570-9571 (2007-07-17)
B Goldfuss et al.
The Journal of organic chemistry, 65(1), 77-82 (2000-05-18)
Three (-)-fenchyl alcohol derivatives, ¿(1R,2R,4S)-exo-(2-Ar)-1,3, 3-trimethylbicyclo[2.2.1] heptan-2-ol, Ar = o-anisyl (2), 2-N-methylimidazolyl (3), 2-N,N-dimethylbenzylamine (4)¿ were synthesized, characterized by X-ray analyses, and employed as precatalysts in diethyl zinc additions to benzaldehyde. Directions and relative degrees of enantioselectivities are rationalized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service