Skip to Content
Merck
All Photos(1)

Key Documents

8.08607

Sigma-Aldrich

Trichloroisocyanuric acid

for synthesis

Synonym(s):

Trichloroisocyanuric acid, 1,3,5-Trichlorohexahydro-1,3,5-triazine-2,4,6-trione, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, TCCA, Bleaching solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl3N3O3
CAS Number:
Molecular Weight:
232.41
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-782-8
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

crystals

potency

406 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

2.0-2.7 (20 °C, 10 g/L in H2O)

mp

225-230 °C (decomposition)

transition temp

flash point >250 °C

density

2.07 g/cm3

bulk density

850 kg/m3

storage temp.

2-30°C

SMILES string

Cl[n]1[c]([n]([c]([n]([c]1=O)Cl)=O)Cl)=O

InChI

1S/C3Cl3N3O3/c4-7-1(10)8(5)3(12)9(6)2(7)11

InChI key

YRIZYWQGELRKNT-UHFFFAOYSA-N

Application


  • Catalyst- and metal-free C(sp2)–H bond selenylation of (N-hetero)-arenes using diselenides and trichloroisocyanuric acid at room temperature: An eco-friendly approach for the C(sp2)–H bond selenylation of imidazopyridines and other N-heteroarenes as well as simple arenes at ambient temperatures (JSS Neto et al., 2023).

  • Organic impurity profiling of 3, 4-methylenedioxyamphetamine (MDA) synthesised from helional: This paper explores the use of trichloroisocyanuric acid in the oxidative rearrangement reaction for synthesizing MDA, presenting an alternative oxidizing agent approach (AL Mercieca et al., 2023).

  • Convenient synthesis of 2-chloro-3-substituted quinoxalines by visible-light-induced radical cascade cyclization of ortho-diisocyanoarenes with sulfinic acids: This research discusses the use of trichloroisocyanuric acid as a Cl-source in visible-light-induced reactions, enhancing the synthesis process (P Natarajan et al., 2024).

  • Synergy between UV light and trichloroisocyanuric acid on methylisothiazolinone degradation: The study presents an advanced oxidation process using ultraviolet and trichloroisocyanuric acid for the degradation of MIT, detailing performance, kinetics, and pathways (Z Chai et al., 2023).


Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service