232106
Phosphorus pentasulfide
99%
Synonym(s):
Phosphorus(V) sulfide
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
P2S5
CAS Number:
Molecular Weight:
222.27
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
514 °C
Recommended Products
Quality Level
Assay
99%
form
powder
bp
514 °C
mp
280-284 °C (lit.)
density
2.09 g/mL at 25 °C (lit.)
SMILES string
S=P(=S)SP(=S)=S
InChI
1S/P2S5/c3-1(4)7-2(5)6
InChI key
HWVAJUNEPOKXLF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Phosphorus pentasulfide is also referred to as phosphorus(V) sulfide. It is widely employed as a thiating, deoxygenating and dehydrating agent in various organic reactions. It readily transforms esters to dithioesters.
Application
Phosphorus pentasulfide may be employed in the preparation of the following:
- Bandgap polymer poly(isothianaphthene).
- 3,5-diiodo-2-thiopyridone and 3,5-diiodo-4-thiopyridone.
- Phosphorus pentasulfide can also be used:,In the conversion of carbonyls to thiocarbonyls.
- To activate alcohols and carboxylic acid.
- To convert furan to thiophene.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Sol. 1 - Water-react 1
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Phosphorus (V) sulfide.
Edmondson SD and Sannigrahi M.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2004)
Thiation with phosphorus pentasulfide in pyridine solution.
Klingsberg E and Papa D.
Journal of the American Chemical Society, 73(10), 4988-4989 (1951)
New synthetic routes to poly (isothianaphthene) I. Reaction of phthalic anhydride and phthalide with phosphorus pentasulfide.
van Asselt R, et al.
Synthetic Metals, 74(1), 65-70 (1995)
Oriane Della-Negra et al.
Scientific reports, 10(1), 13545-13545 (2020-08-13)
The insecticide chlordecone has been used in the French West Indies for decades, resulting in long term pollution, human health problems and social crisis. In addition to bacterial consortia and Citrobacter sp.86 previously described to transform chlordecone into three families
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service