Skip to Content
Merck
All Photos(1)

Key Documents

681665

Sigma-Aldrich

Wnt Agonist

The Wnt Agonist, also referenced under CAS 853220-52-7, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Wnt Agonist, 2-Amino-4-(3,4-(methylenedioxy)benzylamino)-6-(3-methoxyphenyl)pyrimidine, Wnt Pathway Activator I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4O3
CAS Number:
Molecular Weight:
350.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid (sticky)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellowish

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(Cc3cc4c(cc3)OCO4)c1nc(nc(c1)c2cc(ccc2)OC)N

InChI

1S/C19H18N4O3/c1-24-14-4-2-3-13(8-14)15-9-18(23-19(20)22-15)21-10-12-5-6-16-17(7-12)26-11-25-16/h2-9H,10-11H2,1H3,(H3,20,21,22,23)

InChI key

FABQUVYDAXWUQP-UHFFFAOYSA-N

General description

A cell-permeable, potent, and selective activator of Wnt signaling that does not inhibit the activity of GSK-3β (IC50 >60 µM). Shown to mimic the effect of Wnt and induce β-catenin and TCF (T-cell fate)-dependent transcriptional activity (EC50 = 700 nM in HEK-293T cells). Reported to cause substantial head defects during Xenopus embryonic development.

Biochem/physiol Actions

Cell permeable: yes
EC50 = 700 nM inducing β-catenin and TCF (T-cell fate)-dependent transcriptional activity in HEK-293T cells
Primary Target
Selective activator of Wnt signaling
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Liu, J., et al. 2005. Angew. Chem. Int. Ed.44, 1987.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chiharu Kimura-Yoshida et al.
Communications biology, 5(1), 378-378 (2022-04-21)
Previously, we have shown that the translocation of Grainyhead-like 3 (GRHL3) transcription factor from the nucleus to the cytoplasm triggers the switch from canonical Wnt signaling for epidermal differentiation to non-canonical Wnt signaling for epithelial morphogenesis. However, the molecular mechanism
Camille Cohen et al.
Nature communications, 15(1), 743-743 (2024-01-26)
Chronic kidney disease (CKD) is a public health problem driven by myofibroblast accumulation, leading to interstitial fibrosis. Heterogeneity is a recently recognized characteristic in kidney fibroblasts in CKD, but the role of different populations is still unclear. Here, we characterize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service