Merck
  • WNT-dependent interaction between inflammatory fibroblasts and FOLR2+ macrophages promotes fibrosis in chronic kidney disease.
WNT-dependent interaction between inflammatory fibroblasts and FOLR2+ macrophages promotes fibrosis in chronic kidney disease.
Nature communications (2024-01-26)
Camille Cohen, Rana Mhaidly, Hugo Croizer, Yann Kieffer, Renaud Leclere, Anne Vincent-Salomon, Catherine Robley, Dany Anglicheau, Marion Rabant, Aurélie Sannier, Marc-Olivier Timsit, Sean Eddy, Matthias Kretzler, Wenjun Ju, Fatima Mechta-Grigoriou
ABSTRACT
Chronic kidney disease (CKD) is a public health problem driven by myofibroblast accumulation, leading to interstitial fibrosis. Heterogeneity is a recently recognized characteristic in kidney fibroblasts in CKD, but the role of different populations is still unclear. Here, we characterize a proinflammatory fibroblast population (named CXCL-iFibro), which corresponds to an early state of myofibroblast differentiation in CKD. We demonstrate that CXCL-iFibro co-localize with macrophages in the kidney and participate in their attraction, accumulation, and switch into FOLR2+ macrophages from early CKD stages on. In vitro, macrophages promote the switch of CXCL-iFibro into ECM-secreting myofibroblasts through a WNT/β-catenin-dependent pathway, thereby suggesting a reciprocal crosstalk between these populations of fibroblasts and macrophages. Finally, the detection of CXCL-iFibro at early stages of CKD is predictive of poor patient prognosis, which shows that the CXCL-iFibro population is an early player in CKD progression and demonstrates the clinical relevance of our findings.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Wnt Agonist, The Wnt Agonist, also referenced under CAS 853220-52-7, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Anti-RAMP1 Antibody, clone 4F8.1, clone 4F8.1, from mouse
Sigma-Aldrich
β-Catenin/Tcf Inhibitor III, iCRT3, The β-Catenin/Tcf Inhibitor III, iCRT3 controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Biochemicals applications.
Sigma-Aldrich
Wnt Agonist II, SKL2001, The Wnt Agonist II, SKL2001 controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Biochemicals applications.