Skip to Content
Merck
All Photos(3)

Key Documents

I0161

Sigma-Aldrich

IWR-1

≥98% (HPLC), powder, Tankyrase1/2 inhibitor

Synonym(s):

4-(1,3,3a,4,7,7a-Hexahydro-1,3-dioxo-4,7-methano-2H-isoindol-2-yl)-N-8-quinolinyl-Benzamide, IWR1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N3O3
CAS Number:
Molecular Weight:
409.44
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

IWR-1, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(Nc1cccc2cccnc12)c3ccc(cc3)N4C(=O)[C@@H]5[C@H]6C[C@H](C=C6)[C@@H]5C4=O

InChI

1S/C25H19N3O3/c29-23(27-19-5-1-3-14-4-2-12-26-22(14)19)15-8-10-18(11-9-15)28-24(30)20-16-6-7-17(13-16)21(20)25(28)31/h1-12,16-17,20-21H,13H2,(H,27,29)/t16-,17+,20-,21+

InChI key

ZGSXEXBYLJIOGF-ALFLXDJESA-N

Application

IWR-1 has been used as an antagonist for Wingless/Integrated (WNT) signaling pathway in human embryonic stem cells (hESCs), mice lewis lung carcinoma (LLC) and Sepia embryos.

Biochem/physiol Actions

IWR-1 is a tankyrase inhibitor and is cytotoxic for cancer stem-like cells. It inhibits the in vivo progression of osteosarcoma. IWR-1 interacts with RNA polymerase II and is crucial for the formation of preinitiation complex.
The Wnt/b-catenin (‘canonical′) pathway maintains transcriptional programs that enable stem cells to remain multipotent.
The Wnt/b-catenin (‘canonical′) pathway maintains transcriptional programs that enable stem cells to remain multipotent. Hyperactivation of the Wnt/b-catenin pathway leads to disease stage. IWR-3 act as inhibitors of Wnt response. It appear that IWR compounds induce stabilization of Axin proteins via a direct interaction, which is a part of the b-catenin destruction complex (consists of Apc, Axin, Ck1 and Gsk3b).

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iwr1 protein is important for preinitiation complex formation by all three nuclear RNA polymerases in Saccharomyces cerevisiae
Esberg A, et al.
PLoS ONE, 6(6), e20829-e20829 (2011)
Fei Gao et al.
Cancer letters, 402, 177-189 (2017-06-13)
Akt1 is essential for the oncogenic transformation and tumor growth in various cancers. However, the precise role of Akt1 in advanced cancers is conflicting. Using a neuroendocrine TRansgenic Adenocarcinoma of the Mouse Prostate (TRAMP) model, we first show that the
Efficient differentiation of TBX18+/WT1+ epicardial-like cells from human pluripotent stem cells using small molecular compounds
Zhao J, et al.
Stem Cells and Development, 26(7), 528-540 (2017)
Hirokazu Narita et al.
Scientific reports, 7(1), 13708-13708 (2017-10-24)
One of the major challenges in cell-based cardiac regenerative medicine is the in vitro construction of three-dimensional (3D) tissues consisting of induced pluripotent stem cell-derived cardiomyocyte (iPSC-CM) and a blood vascular network supplying nutrients and oxygen throughout the tissue after
A serine in exon 11 determines the full transcriptional activity of TCF-4 in lung carcinoma cells
Yang B, et al.
Biochemical and biophysical research communications, 508(3), 675-681 (2019)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service