Skip to Content
Merck
All Photos(2)

Documents

569397

Sigma-Aldrich

Staurosporine

from Streptomyces sp., ≥99% (HPLC), solid, protein kinase inhibitor, Calbiochem

Synonym(s):

Staurosporine, Streptomyces sp., PKA Inhibitor II, MLCK Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Staurosporine, Streptomyces sp., Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC₅₀ = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C28H26N4O3/c1-28-26(34-3)17(29-2)12-20(35-28)31-18-10-6-4-8-14(18)22-23-16(13-30-27(23)33)21-15-9-5-7-11-19(15)32(28)25(21)24(22)31/h4-11,17,20,26,29H,12-13H2,1-3H3,(H,30,33)/t17-,20-,26-,28+/m1/s1

InChI key

HKSZLNNOFSGOKW-FYTWVXJKSA-N

General description

A potent, cell-permeable, reversible, ATP-competitive and broad spectrum inhibitor of protein kinases. Inhibits CaM kinase (IC50 = 20 nM), myosin light chain kinase (IC50 = 1.3 nM), protein kinase A (IC50 = 7 nM), protein kinase C (IC50 = 0.7 nM), and protein kinase G (IC50 = 8.5 nM). Also inhibits platelet aggregation induced by collagen or ADP but has no effect on thrombin-induced platelet aggregation. Induces apoptosis in human malignant glioma cell lines. Arrests normal cells at the G1 checkpoint.
A potent, cell-permeable, reversible, ATP-competitive and broad spectrum inhibitor of protein kinases. Inhibits protein kinase A (IC50 = 7 nM), CaM kinase (IC50 = 20 nM), myosin light chain kinase (IC50 = 1.3 nM), protein kinase C (IC50 = 700 pM), and protein kinase G (IC50 = 8.5 nM). Also inhibits platelet aggregation induced by collagen or ADP but has no effect on thrombin-induced platelet aggregation. Induces apoptosis in human malignant glioma cell lines. Arrests normal cells at the G1 checkpoint. A 1 mM (100 µg/214 µl) solution of Staurosporine (Cat. No. 569396) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKA
Product competes with ATP.
Reversible: yes
Target IC50: 7 nM, 20 nM, 1.3 nM, 700 pM, and 8.5 nM against protein kinase A , CaM kinase, myosin light chain kinase, protein kinase C, and protein kinase G, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.

Other Notes

Couldwell, W.T., et al. 1994. FEBS Lett.345, 43.
Nishimura, H. and Simpson, I.A. 1994. Biochem. J. 302, 271.
Bruno, S., et al. 1992. Cancer Res.52, 470.
Kiss, Z. and Deli, E. 1992. Biochem. J. 288, 853.
Vitale, M.L., et al. 1992. Neuroscience51, 463.
Hoffman, R. and Newland, E.S. 1991. Cancer Chemother. Pharmacol.28, 102.
Oka, S., et al. 1986. Agric. Biol. Chem.50, 2723.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Martínez-Escardó et al.
Cancers, 13(21) (2021-11-14)
Glioblastoma (GBM) is a highly aggressive brain tumor and almost all patients die because of relapses. GBM-derived cells undergo cell death without nuclear fragmentation upon treatment with different apoptotic agents. Nuclear dismantling determines the point-of-no-return in the apoptotic process. DFF40/CAD
Aneese A Jaffa et al.
Frontiers in pharmacology, 12, 743059-743059 (2021-12-07)
Microglia, the resident phagocytes of the central nervous system and one of the key modulators of the innate immune system, have been shown to play a major role in brain insults. Upon activation in response to neuroinflammation, microglia promote the
Alain Pulfer et al.
eLife, 12 (2024-03-18)
Intravital microscopy has revolutionized live-cell imaging by allowing the study of spatial-temporal cell dynamics in living animals. However, the complexity of the data generated by this technology has limited the development of effective computational tools to identify and quantify cell
Wenjia Xie et al.
Cell death & disease, 14(10), 663-663 (2023-10-11)
An important pathophysiological process of acute kidney injury (AKI) is mitochondrial fragmentation in renal tubular epithelial cells, which leads to cell death. Pyruvate kinase M2 (PKM2) is an active protein with various biological functions that participates in regulating glycolysis and
Tomoko Funakoshi et al.
Cell structure and function, 49(2), 31-46 (2024-06-06)
In metazoans, the nuclear envelope (NE) disassembles during the prophase and reassembles around segregated chromatids during the telophase. The process of NE formation has been extensively studied using live-cell imaging. At the early step of NE reassembly in human cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service