Skip to Content
Merck
All Photos(1)

Key Documents

M1555

Sigma-Aldrich

MAHMA NONOate

Synonym(s):

6-(2-Hydroxy-1-methyl-2-nitrosohydrazino)-N-methyl-1-hexanamine, NOC-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20N4O2
CAS Number:
Molecular Weight:
204.27
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

shipped in

dry ice

storage temp.

−70°C

SMILES string

[H][N+]([H])(C)CCCCCCN(C)N([O-])N=O

InChI

1S/C8H19N4O2/c1-9-7-5-3-4-6-8-11(2)12(14)10-13/h9H,3-8H2,1-2H3/q-1/p+1

InChI key

WASRCWNCCFPNEJ-UHFFFAOYSA-O

Application

NO donor

Biochem/physiol Actions

NO release: t1/2 = 3 min (phosphate buffer at 22° C)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Muenster et al.
Anesthesiology, 125(5), 952-963 (2016-10-19)
Transfusion of packed erythrocytes stored for a long duration is associated with increased pulmonary arterial pressure and vascular resistance. Prolonged storage decreases erythrocyte deformability, and older erythrocytes are rapidly removed from the circulation after transfusion. The authors studied whether treating
H P Monteiro et al.
Free radical biology & medicine, 28(2), 174-182 (2001-04-03)
Nitric oxide (NO) can participate in cellular signaling. In this study, monoclonal antibodies against proteins from the growth factor-mediated signalling pathway were used to identify a set of 126-, 56-, 43-, and 40-kDa proteins phosphorylated on tyrosine at NO stimulation
D Clark et al.
Molecular plant-microbe interactions : MPMI, 13(12), 1380-1384 (2000-12-06)
We used a variety of nitric oxide (NO) donors to demonstrate that NO inhibits the activities of tobacco catalase and ascorbate peroxidase (APX). This inhibition appears to be reversible because removal of the NO donor led to a significant recovery
S H Lin et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 20(9), 1380-1391 (2000-09-20)
Neuronal injury may be dependent upon the generation of the free radical nitric oxide (NO) and the subsequent induction of programmed cell death (PCD). Although the nature of this injury may be both preventable and reversible, the underlying mechanisms that
A M Vincent et al.
Journal of neurobiology, 40(2), 171-184 (1999-07-21)
Neuronal injury is intricately linked to the activation of three distinct neuronal endonucleases. Since these endonucleases are exquisitely pH dependent, we investigated in primary rat hippocampal neurons the role of intracellular pH (pH(i)) regulation during nitric oxide (NO)-induced toxicity. Neuronal

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service