Skip to Content
Merck
All Photos(2)

Key Documents

E1383

Sigma-Aldrich

Etoposide

95.0-105.0% (anhydrous basis), powder, topoisomerase II inhibitor

Synonym(s):

4′-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), VP-16-213

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32O13
CAS Number:
Molecular Weight:
588.56
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Etoposide, synthetic, 95.0-105.0%, powder

biological source

synthetic

Assay

95.0-105.0%

form

powder

color

white to off-white

pKa 

9.8

mp

236-251 °C (lit.)

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes
enzyme | inhibits

originator

Teva

SMILES string

COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@@H]5CO[C@@H](C)O[C@H]5[C@H](O)[C@H]4O)c6cc7OCOc7cc26

InChI

1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3/t11-,15+,20-,21-,22+,24-,25-,26-,27-,29+/m1/s1

InChI key

VJJPUSNTGOMMGY-MRVIYFEKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Etoposide belongs to the class of medications known as topoisomerase II inhibitors. This semi-synthetic compound is derived from Podophyllum peltatum, and Podophyllum emodi. The complex formed between etoposide and topoisomerase II initiates a mutagenic and cell death pathway, demonstrating optimal efficacy in tumor cells with elevated levels of topoisomerase II enzymes. Etoposide is administered as an oral or intravenous chemotherapy agent.
Etoposide is synthesised from podophyllotoxins of plants.

Application

Etoposide has been used:
  • to prepare drug stock solution in dimethyl sulfoxide (DMSO) and also to profile and compare the sensitivity of DT40 mutant cells
  • to incubate cells for cell viability assay
  • to treat neuro-2A cells to induce programmed cell death

Biochem/physiol Actions

Antitumor agent that complexes with topoisomerase II and DNA to enhance double-strand and single-strand cleavage of DNA and reversibly inhibit religation.
Etoposide is an antitumor agent that complexes with topoisomerase II and DNA to enhance double-strand and single-strand cleavage of DNA and reversibly inhibit religation. Blocks the cell cycle in in S-phase and G2-phase of the cell cycle; induces apoptosis in normal and tumor cell lines; inhibits synthesis of the oncoprotein Mdm2 and induces apoptosis in tumor lines that overexpress Mdm2.
Etoposide is used in treating advanced testicular cancer, Kaposi′s sarcoma, non-small cell lung cancer (NSCLC), SCLC (small cell lung cancer) and lymphoma.

Features and Benefits

This compound is a featured product for ADME Tox and Apoptosis research. Discover more featured ADME Tox and Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Teva. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential and common DNA repair pathways for topoisomerase I-and II-targeted drugs in a genetic DT40 repair cell screen panel
Maede Y, et al.
Molecular Cancer Therapeutics, 13(1), 214-220 (2014)
Etoposide
Reyhanoglu G and Tadi P
StatPearls [Internet] (2023)
The latency-related gene of bovine herpesvirus 1 inhibits programmed cell death
Ciacci-Zanella J, et al.
Journal of Virology, 73(12), 9734-9740 (1999)
Pharmacology and Therapeutics for Dentistry - E-Book, 700-700 (2010)
Loss of TBL1XR1 disrupts glucocorticoid receptor recruitment to chromatin and results in glucocorticoid resistance in a B-lymphoblastic leukemia model
Jones CL, et al.
Test, jbc-M114 (2014)

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service