Skip to Content
Merck
All Photos(1)

Key Documents

309800

Sigma-Aldrich

Dihydroethidium

A cell-permeable, chemically-reduced ethidium derivative.

Synonym(s):

Dihydroethidium, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine, DHE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3
CAS Number:
Molecular Weight:
315.41
MDL number:
UNSPSC Code:
12161505
NACRES:
NA.25

Quality Level

description

RTECS - SF7935000

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pink

solubility

DMSO: 20 mg/mL
DMF: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

Nc1c2c(cnc1)c3c(cc(cc3)N)C(C2CC)c4ccccc4

InChI

1S/C21H21N3/c1-2-15-20(13-6-4-3-5-7-13)17-10-14(22)8-9-16(17)18-11-24-12-19(23)21(15)18/h3-12,15,20H,2,22-23H2,1H3

InChI key

ZDWPSSBKIMIPMY-UHFFFAOYSA-N

General description

A cell-permeable, chemically reduced ethidium derivative that can be used as a fluorogenic probe for the detection of reactive oxygen species (ROS). It undergoes significant oxidation in resting leukocytes, possibly through the uncoupling of mitochondrial oxidative phosphorylation. Cytosolic dihydroethidium displays blue fluorescence, however, following oxidation to ethidium and subsequent intercalation to DNA, a bright red fluorescence is achieved. Often used to determine the level of superoxide anion in cells by flow cytometry.
A cell-permeable, chemically-reduced ethidium derivative. Commonly used to analyze respiratory burst in phagocytes. Useful fluorogenic probe for the detection of reactive oxygen species (ROS). It undergoes significant oxidation in resting leukocytes, possibly through the uncoupling of mitochondrial oxidative phosphorylation. Cytosolic dihydroethidium displays blue fluorescence, whereas after oxidation of this probe to ethidium, it intercalates cell’s DNA, staining the nucleus with a bright red fluorescence.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Useful fluorogenic probe for the detection of reactive oxygen species (ROS)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot, flush with an inert gas, and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Haugen, T.S., et al. 1999. Eur. Respir. J.14, 1100.
Mancini, M., et al. 1997. J. Cell Biol.138, 449.
Carter, W.O., et al. 1994. J. Leukoc. Biol.55, 253.
Cinco, M., et al. 1994. FEMS Microbiol. Lett. 122, 187.
Perticarari, S., et al. 1994. J. Immunol. Methods 170, 117.
Perticarari, S., et al. 1991. Cytometry 12, 687.
Bucana, C.D., et al. 1990. Exp. Cell Res.190, 69.
Rothe, G., and Valet, G. 1990. J. Leukoc. Biol.47, 440.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service