309825
Dihydrorhodamine 123
Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) such as peroxide and peroxynitrite.
Synonym(s):
Dihydrorhodamine 123
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pink-white
solubility
DMSO: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
Nc1cc2c(cc1)C(c4c(cc(cc4)N)O2)c3c(cccc3)C(=O)OC
InChI
1S/C21H18N2O3/c1-25-21(24)15-5-3-2-4-14(15)20-16-8-6-12(22)10-18(16)26-19-11-13(23)7-9-17(19)20/h2-11,20H,22-23H2,1H3
InChI key
FNEZBBILNYNQGC-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Detection of reactive oxygen species (ROS)
Packaging
Warning
Reconstitution
Other Notes
Kim, Y.M., et al. 1996. Infect Immun. 64, 3074.
Williams, M.S., and Henkart, P.A. 1996. J. Immunol. 157, 2395.
van de Water, B., et al. 1995. Mol. Pharmacol. 48, 928.
Royall, J.A., and Ischiropoulos, H. 1993. Arch. Biochem. Biophys. 302, 348.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service