Skip to Content
Merck
All Photos(1)

Key Documents

300260

Sigma-Aldrich

Diphenyleneiodonium Chloride

≥97% (HPLC), crystalline solid, eNOS inhibitor, Calbiochem®

Synonym(s):

Diphenyleneiodonium Chloride, DPI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8ClI
CAS Number:
Molecular Weight:
314.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Diphenyleneiodonium Chloride, A cell-permeable, irreversible inhibitor of endothelial nitric oxide synthase (eNOS).

Quality Level

Assay

≥97% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[I+]1c2c(cccc2)c3c1cccc3.[Cl-]

InChI

1S/C12H8I.ClH/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11;/h1-8H;1H/q+1;/p-1

InChI key

FCFZKAVCDNTYID-UHFFFAOYSA-M

General description

A cell-permeable, irreversible endothelial nitric oxide synthase inhibitor. Inhibits iNOS in macrophages (IC50 = 5 nM). Shown to inhibit acetylcholine-induced relaxation of preconstricted rat thoracic aortic rings (IC50 = 180 nM). Blocks K+ and Ca2+ currents in arterial smooth muscle cells. Inhibitor of mitochondrial NADH-ubiquinone oxidoreductase. Inhibits other NADH utilizing flavoproteins, such as NADPH oxidase in human neutrophils and macrophages. Causes pressor and tachycardiac effects via indirect activation of the sympathetic nervous system.
A cell-permeable, irreversible inhibitor of endothelial nitric oxide synthase (eNOS). Shown to inhibit acetylcholine-induced relaxation of preconstricted rat thoracic aortic rings (IC50 = 180 nM). Also inhibits iNOS in macrophages (IC50 = 50 nM). Blocks K+ and Ca2+ currents in arterial smooth muscle cells. Inhibitor of mitochondrial NADPH-ubiquinone oxido-reductase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
eNOS
Product does not compete with ATP.
Reversible: no
Target IC50: 180 nM against endothelial nitric oxide synthase (eNOS)

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Preparation Note

Slight warming is required for complete solubility.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, Y.X., et al. 1995. Br. J. Pharmacol.114, 194.
Majander, A., et al. 1994. J. Biol. Chem. 269, 21037.
Weir, E.K., et al. 1994. J. Appl. Physiol. 76, 2611.
Wang, Y.X., et al. 1993. Br. J. Pharmacol.110, 1232.
Wang, Y.X., and Pang, C.C. 1993. Br. J. Pharmacol.109, 1186.
Stuehr, D.J., et al. 1991. FASEB J.5, 98.
Handcock, J.T., and Jones, O.T.G. 1987. Biochem. J.242, 103.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Pereira Gonçalves et al.
iScience, 27(3), 109248-109248 (2024-03-04)
Mesenchymal stromal cells (MSCs) have been modified via genetic or pharmacological engineering into potent antigen-presenting cells-like capable of priming responding CD8 T cells. In this study, our screening of a variant library of Accum molecule revealed a molecule (A1) capable of
Omar Rafael Alemán et al.
Frontiers in immunology, 12, 657393-657393 (2021-06-01)
Human neutrophils express two unique antibody receptors for IgG, the FcγRIIa and the FcγRIIIb. FcγRIIa contains an immunoreceptor tyrosine-based activation motif (ITAM) sequence within its cytoplasmic tail, which is important for initiating signaling. In contrast, FcγRIIIb is a glycosylphosphatidylinositol (GPI)-linked
Saurav Kumar et al.
eLife, 12 (2023-07-25)
Integrin-mediated cell attachment rapidly induces tyrosine kinase signaling. Despite years of research, the role of this signaling in integrin activation and focal adhesion assembly is unclear. We provide evidence that the Src-family kinase (SFK) substrate Cas (Crk-associated substrate, p130Cas, BCAR1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service