Skip to Content
Merck
All Photos(1)

Key Documents

178385

Sigma-Aldrich

Apocynin

≥98% (GC), powder, NADPH oxidase inhibitor, Calbiochem®

Synonym(s):

Apocynin, 4-Hydroxy-3-methoxyacetophenone, Acetovanillone, NADPH Oxidase Inhibitor V, NOX Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Apocynin, A cell-permeable, anti-inflammatory phenolic compound that acts as a potent and selective inhibitor of NADPH oxidase.

Quality Level

description

Merck USA index - 14, 741

Assay

≥98% (GC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 10 mg/mL
ethanol: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O(C)c1c(ccc(c1)C(=O)C)O

InChI

1S/C9H10O3/c1-6(10)7-3-4-8(11)9(5-7)12-2/h3-5,11H,1-2H3

InChI key

DFYRUELUNQRZTB-UHFFFAOYSA-N

General description

A cell-permeable, anti-inflammatory phenolic compound that acts as a potent and selective inhibitor of NADPH oxidase. Shown to block peroxynitrite formation in murine macrophages. Reported to increase glutathione synthesis through activation of AP-1. Also reported to prevent phagocytosis of myelin by macrophages (10 mM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NADPH oxidase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Abid, M.R., et al. 2000. FEBS Lett.486, 252.
Muijsers, R.B., et al. 2000. Br. J. Pharmacol.130, 932.
Lapperre, T.S., et al. 1999. FEBS Lett.443, 235.
Supinski, G., et al. 1999, J. Appl. Physiol.87, 776.
van der Goes, A., et al. 1998. J. Neuroimmunol.92, 67.
Hart, B.A., and Simmons, J.M. 1992. Biotechnol. Ther.3, 3.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nguyen Mai et al.
Journal of neurochemistry, 168(5), 899-909 (2024-02-01)
Cofilactin rods (CARs), which are 1:1 aggregates of cofilin-1 and actin, lead to neurite loss in ischemic stroke and other disorders. The biochemical pathways driving CAR formation are well-established, but how these pathways are engaged under ischemic conditions is less
Nagat Frara et al.
PloS one, 18(6), e0287212-e0287212 (2023-06-23)
Roles of redox signaling in bladder function is still under investigation. We explored the physiological role of reactive oxygen species (ROS) and nicotinamide adenine dinucleotide phosphate (NADPH) oxidase (Nox) in regulating bladder function in humans and dogs. Mucosa-denuded bladder smooth
Yangyang Ding et al.
Nature communications, 14(1), 4829-4829 (2023-08-11)
Human MutT Homolog 1 (MTH1) is a nucleotide pool sanitization enzyme that hydrolyzes oxidized nucleotides to prevent their mis-incorporation into DNA under oxidative stress. Expression and functional roles of MTH1 in platelets are not known. Here, we show MTH1 expression
Kendra M Prutton et al.
Free radical biology & medicine, 196, 11-21 (2023-01-14)
Cellular redox state directs differentiation of induced pluripotent stem cells (iPSC) by energy metabolism control and ROS generation. As oxidative stress and mitochondrial dysfunction have been extensively reported in Down syndrome (DS), we evaluated mitochondrial phenotypes and energy metabolism during
Ji-Hee Kim et al.
International journal of molecular sciences, 25(11) (2024-06-19)
Urban air pollution, a significant environmental hazard, is linked to adverse health outcomes and increased mortality across various diseases. This study investigates the neurotoxic effects of particulate matter (PM), specifically PM2.5 and PM10, by examining their role in inducing oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service