Skip to Content
Merck

Skip To

P3053

Penitrem A

≥95% (HPLC and TLC)

Synonym(s):

Tremortin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C37H44ClNO6
CAS Number:
Molecular Weight:
634.20
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC and TLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC and TLC)

form

powder

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@@]3(C)[C@@](O)(CC[C@@]4([H])[C@]5([H])OC(C)(C)[C@@]6([H])C[C@]7([H])C(=C)Cc8c(Cl)cc9[nH]c(c5c9c8[C@]67O)[C@]34C)[C@]1%10O[C@]%10([H])[C@@H](O)[C@H](O2)C(C)=C

InChI

1S/C37H44ClNO6/c1-15(2)28-27(40)31-37(45-31)23(43-28)9-10-33(6)34(7)18(8-11-35(33,37)41)29-25-24-21(39-30(25)34)14-20(38)17-12-16(3)19-13-22(32(4,5)44-29)36(19,42)26(17)24/h14,18-19,22-23,27-29,31,39-42H,1,3,8-13H2,2,4-7H3/t18-,19+,22+,23-,27-,28+,29-,31+,33+,34+,35-,36+,37-/m0/s1

InChI key

JDUWHZOLEDOQSR-JKPSMKLGSA-N

Application

Penitrem A has been used as Ca2+-Activated K+ (BK) channel blocker to study its effect on BK current in bag cell neurons.

Biochem/physiol Actions

Penitrem A intoxication causes ataxia, polypnea, and sustained tremors and may lead to seizures and death. Penitrem A affects the central and peripheral nervous system. It impairs the GABAergic neurotransmission in the cerebellum.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Originally isolated from Penicillium palitans

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1584W1628N7143
assay

≥95% (HPLC and TLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC and TLC)

assay

≥98% (TLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

100

Quality Level

300

Quality Level

300

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters