Skip to Content
Merck
All Photos(1)

Documents

1.01543

Millipore

L-Arginine monohydrochloride

for biochemistry

Synonym(s):

α-Amino-δ-guanidino valerianic acid hydrochloride, alpha-Amino-delta-guanidino valerianic acid hydrochloride, Arg, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
MDL number:
UNSPSC Code:
12352209
EC Index Number:
214-275-1
NACRES:
NA.21

vapor pressure

<1 Pa ( 20 °C)

Quality Level

Assay

99% dry basis (perchloric acid titration)

form

powder

potency

12000 mg/kg LD50, oral (Rat)

pH

5.7 (20 °C, 100 g/L in H2O)

mp

>235 °C

solubility

730 g/L

density

1.42 g/cm3 at 20 °C

bulk density

1250 kg/m3

anion traces

sulfate (SO42-): ≤0.01%

cation traces

As: ≤0.0005%
Ca: ≤0.001%
Co: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.01%
Na: ≤0.005%
Zn: ≤0.0005%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Improvement in Dental Health Products: Studies have shown that L-Arginine monohydrochloride enhances the bioavailability of fluoride in dental care products, promoting better oral health (Bijle et al., 2020).

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine powder
Appearance of solution (1 mol/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +21.0 to +22.0
UV-Absorption(260 nm; 1 mol/l; 1 cm; water): ≤ 0.20
UV-Absorption(280 nm; 1 mol/l; 1 cm; water): ≤ 0.05
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Zn (Zinc): ≤ 0.0005 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.2 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service