Skip to Content
Merck
All Photos(2)

Documents

A4599

SAFC

L-Arginine monohydrochloride

Pharma Manufacturing

Synonym(s):

(S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

biological source

non-animal source

Quality Level

Assay

≥98.5%

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, heavy metals, residual solvents, tested

solubility

H2O: 100 mg/mL

density

1.42 g/cm3 at 20 °C

anion traces

chloride (Cl-): 16.58-17.00%
sulfate (SO42-): ≤0.020%

cation traces

As: ≤1.0 ppm
NH4+: ≤0.02%
heavy metals: ≤10 ppm

application(s)

pharmaceutical (small molecule)

foreign activity

cytotoxicity, tested

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.

Application

L-Arginine is a non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-Arginine monohydrochloride is a modified version of L-Arginine with improved solubility.

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Canobbio et al.
The Biochemical journal, 462(3), 513-523 (2014-07-02)
Alzheimer's disease is associated with the accumulation of Aβ (amyloid β)-peptides in the brain. Besides their cytotoxic effect on neurons, Aβ-peptides are thought to be responsible for the atherothrombotic complications associated with Alzheimer's disease, which are collectively known as cerebrovascular
Carolina Di Somma et al.
Endocrine, 49(2), 492-502 (2014-11-10)
The aim of the study is to clarify the relationship between adipose tissue dysfunction, metabolic profile and growth hormone (GH)/insulin-like growth factor (IGF)-I secretion in healthy adult subjects. We investigated the metabolic profile in a cohort of 231 consecutive healthy
C B Christiansen et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 47(7), 537-543 (2015-01-21)
TLQP-21 is a VGF-derived neuropeptide proposed to be involved in regulation of metabolism. More specifically it has been suggested that TLQP-21 has the ability to enhance glucose stimulated insulin secretion, making it a candidate for treatment of patients with type
Nivaldo Ribeiro Villela et al.
Journal of translational medicine, 12, 232-232 (2014-08-26)
Relative hypovolemia is frequently found in early stages of severe sepsis and septic shock and prompt and aggressive fluid therapy has become standard of care improving tissue perfusion and patient outcome. This paper investigates the role of the nitric oxide
Jenna Gallie et al.
PLoS biology, 13(3), e1002109-e1002109 (2015-03-13)
Phenotype switching is commonly observed in nature. This prevalence has allowed the elucidation of a number of underlying molecular mechanisms. However, little is known about how phenotypic switches arise and function in their early evolutionary stages. The first opportunity to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service