Skip to Content
Merck
All Photos(1)

Key Documents

860686P

Avanti

15:0 SM(d18:1/15:0)-d9

Avanti Research - A Croda Brand (860686P), powder

Synonym(s):

N-pentadecanoyl-D-erythro-sphingosylphosphorylcholine-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H68D9N2O6P
CAS Number:
Molecular Weight:
698.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (330727P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand (860686P)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCC)=O)COP([O-])(OCC[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O

InChI

1S/C38H77N2O6P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-37(41)36(35-46-47(43,44)45-34-33-40(3,4)5)39-38(42)32-30-28-26-24-22-19-17-15-13-11-9-7-2/h29,31,36-37,41H,6-28,30,32-35H2,1-5H3,(H-,39,42,43,44)/b31-29+/t36-,37+/m0/s1/i3D3,4D3,5D3

General description

15:0 SM(d18:1/15:0)-d9 is a deuterated derivative of 15:0 SM(d18:1/15:0). SM(d18:1/15:0) is composed of a pentadecanoic acid chain and a sphingosine backbone. Sphingomyelin is a major phospholipid in the plasma membrane of eukaryotic cells. It is found at higher concentrations in brain, ocular lenses and peripheral nervous tissues.

Biochem/physiol Actions

Sphingomyelin (SM) regulates the cholesterol distribution and cholesterol homeostasis in cells. It is a precursor of sphingolipid signaling molecules.

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The functional role of sphingomyelin in cell membranes
Slotte J P and Ramstedt B
European Journal of Lipid Science and Technology, 109(10), 977-981 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service