Skip to Content
Merck
All Photos(2)

Key Documents

700041P

Avanti

cholesterol-d7

Avanti Research - A Croda Brand

Synonym(s):

cholesterol-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39OD7
CAS Number:
Molecular Weight:
393.70
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (700041P-10mg)
pkg of 1 × 200 mg (700041P-200mg)
pkg of 1 × 25 mg (700041P-25mg)
pkg of 1 × 50 mg (700041P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3,18D

InChI key

HVYWMOMLDIMFJA-IFAPJKRJSA-N

General description

Cholesterol-d7 is a deuterium labeled cholesterol. It forms D6-steroidal glycoalkaloids (SGA).[1]

Application


  • Quantification of Endogenous Cholesterol in Human Serum on Paper Using Direct Analysis in Real Time Mass Spectrometry.: This study describes a novel technique using deuterium-labeled cholesterol for the quantification of endogenous cholesterol in human serum, offering advancements in metabolic profiling and diagnostics (Hsieh et al., 2017).

Packaging

5 mL Amber Glass Screw Cap Vial (700041P-10mg)
5 mL Amber Glass Screw Cap Vial (700041P-200mg)
5 mL Amber Glass Screw Cap Vial (700041P-25mg)
5 mL Amber Glass Screw Cap Vial (700041P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conversion of exogenous cholesterol into glycoalkaloids in potato shoots, using two methods for sterol solubilisation
Petersson EV, et al.
PLoS ONE, 8(12), e82955-e82955 (2013)
Matias M Vazquez et al.
Redox biology, 36, 101591-101591 (2020-06-13)
Macrophages play a pivotal role in the early stages of atherosclerosis development; they excessively accumulate cholesterol in the cytosol in response to modified Low Density Lipoprotein (mLDL). The mLDL are incorporated through scavenger receptors. CD36 is a high-affinity cell surface
Bingpeng Yan et al.
International journal of molecular sciences, 20(23) (2019-11-30)
Enterovirus A71 (EV-A71) and coxsackievirus A16 (CV-A16) are the most common causes of hand, foot, and mouth disease. Severe EV-A71 and CV-A16 infections may be associated with life-threatening complications. However, the pathogenic mechanisms underlying these severe clinical and pathological features
David B Iaea et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(4), 158609-158609 (2020-01-10)
STARD4, a member of the evolutionarily conserved START gene family, is a soluble sterol transport protein implicated in cholesterol sensing and maintenance of cellular homeostasis. STARD4 is widely expressed and has been shown to transfer sterol between liposomes as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service