Skip to Content
Merck
All Photos(2)

Documents

850475P

Avanti

18:1-16:0 PC

Avanti Research - A Croda Brand

Synonym(s):

1--(9Z-octadecenoyl)-2-hexadecanoyl-sn-glycero-3-phosphocholine; OPPC; PC(18:1(9Z)/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (850475P-200mg)
pkg of 1 × 25 mg (850475P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43(3,4)5)51-42(45)35-33-31-29-27-25-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1

InChI key

RRVPPYNAZJRZFR-VYOBOKEXSA-N

Application

18:1-16:0 PC (1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine) may be used:
  • as a lipid standard in ultra-performance liquid chromatography (UPLC) analysis
  • as a component in liposomes to study the cell proliferation of several cell lines
  • in liposomes for cross-linking experiment(5)

Biochem/physiol Actions

Phosphatidylcholine (PC) can form a hydrophobic surface in the mucus by acting as a surfactant to inhibit the penetrance of bacteria.

Packaging

5 mL Clear Glass Sealed Ampule (850475P-200mg)
5 mL Clear Glass Sealed Ampule (850475P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Anna Werlein et al.
Biochimica et biophysica acta, 1848(7), 1599-1608 (2015-04-29)
According to early experiments with natural extracts, phosphatidylcholines (PCs) are widely considered essentially non-toxic. In addition to these physiological mixed-chain PCs, many different synthetic diacyl-PCs are currently available, but they have never been systematically evaluated for any interference with cell
Eun A Ra et al.
Nature communications, 7, 11726-11726 (2016-05-25)
Autophagy is responsible for the bulk degradation of cytosolic constituents and plays an essential role in the intestinal epithelium by controlling beneficial host-bacterial relationships. Atg5 and Atg7 are thought to be critical for autophagy. However, Atg5- or Atg7-deficient cells still
Katharina Wozny et al.
Analytical and bioanalytical chemistry, 411(4), 915-924 (2018-12-24)
Diacyl glycerophospholipids (GPs) belong to the most abundant lipid species in living organisms and consist of a glycerol backbone with fatty acyl groups in sn-1 and sn-2 and a polar head group in the sn-3 position. Regioisomeric mixed diacyl GPs
Wei He et al.
Nanomedicine : nanotechnology, biology, and medicine, 24, 102154-102154 (2020-01-27)
In vivo delivery of large RNA molecules has significant implications for novel gene therapy, biologics delivery, and vaccine applications. We have developed cationic nanolipoprotein particles (NLPs) to enhance the complexation and delivery of large self-amplifying mRNAs (replicons) in vivo. NLPs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service