Skip to Content
Merck
All Photos(2)

Documents

850366C

Avanti

18:0 (9,10dibromo) PC

1,2-di-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(9,10-dibromooctadecanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8PBr4
CAS Number:
Molecular Weight:
1105.73
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850366C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850366C

concentration

10 mg/mL (850366C-10mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:0 (9,10dibromo) PC (1,2-di-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine) is a zwitterionic brominated phospholipid.

Application

18:0 (9,10dibromo) PC (1,2-di-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine) may be used:
  • in the preparation of liposomes/small unilamellar vesicles (SUVs)
  • in the preparation of large unilamellar vesicles (LUVs) to facilitate vesicles sedimentation
  • as a component in liposomes

Biochem/physiol Actions

Phosphatidylcholine (PC) forms a hydrophobic surface in the mucus by acting as a surfactant resulting in the inhibition of penetrance of bacteria.

Packaging

5 mL Clear Glass Sealed Ampule (850366C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Olatz Landeta et al.
The Journal of biological chemistry, 286(10), 8213-8230 (2011-01-05)
BAK is a key effector of mitochondrial outer membrane permeabilization (MOMP) whose molecular mechanism of action remains to be fully dissected in intact cells, mainly due to the inherent complexity of the intracellular apoptotic machinery. Here we show that the
Maria Jesús Sánchez-Martín et al.
Journal of colloid and interface science, 360(1), 124-131 (2011-05-14)
The aim of this study was to identify proteins that could inhibit the activity of the peptide sequence representing the N-terminal of the surface protein gp41 of HIV, corresponding to the fusion peptide of the virus (HIV-1 FP). To do
Jocelyn Méré et al.
The Biochemical journal, 386(Pt 1), 47-56 (2004-11-06)
Gelsolin is a calcium-, pH- and lipid-dependent actin filament severing/capping protein whose main function is to regulate the assembly state of the actin cytoskeleton. Gelsolin is associated with membranes in cells, and it is generally assumed that this interaction is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service