Skip to Content
Merck
All Photos(2)

Key Documents

810150C

Avanti

18:1 Liss Rhod PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H109N4O14PS2
CAS Number:
Molecular Weight:
1301.72
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810150C-1mg)
pkg of 5 × 1 mL (810150C-5mg)
pkg of 5 × 2 mL (810150C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (810150C-10mg)
1 mg/mL (810150C-1mg)
1 mg/mL (810150C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

L-α-Phosphatidylethanolamine (PE) is the second most abundant phospholipid in animals, plants and yeast.

Application

18:1 Liss Rhod PE suitable to be used for labelling DOPC (1,2-dioleoyl-sn-glycero-3-phosphocholine) for visualization purpose.

Biochem/physiol Actions

18:1 Liss Rhod PE is a fluorescent labelled lipid, useful in labelling giant unilamellar vesicles (GUVs). L-α-Phosphatidylethanolamine maintains maembrane integrity.

Packaging

5 mL Amber Glass Screw Cap Vial (810150C-10mg)
5 mL Amber Glass Screw Cap Vial (810150C-1mg)
5 mL Amber Glass Screw Cap Vial (810150C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatiotemporal control of coacervate formation within liposomes
Deshpande S, et al.
Nature Communications, 10(1), 1800-1800 (2019)
A new class of synthetic retinoid antibiotics effective against bacterial persisters
Kim W, et al.
Nature, 556(7699), 103-103 (2018)
Stabilization of liposomes against stress using polyelectrolytes: Interaction mechanisms, influence of pH, molecular weight, and polyelectrolyte structure
Rinaudo M, et al.
International Journal of Polymer Analysis and Characterization, 14(8), 667-677 (2009)
Yang Liu et al.
Nature communications, 10(1), 5108-5108 (2019-11-11)
Mounting evidence suggests that the tumor microenvironment is profoundly immunosuppressive. Thus, mitigating tumor immunosuppression is crucial for inducing sustained antitumor immunity. Whereas previous studies involved intratumoral injection, we report here an inhalable nanoparticle-immunotherapy system targeting pulmonary antigen presenting cells (APCs)
Yachong Guo et al.
ACS nano (2018-11-20)
Increasing awareness of bioeffects and toxicity of nanomaterials interacting with cells puts in focus the mechanisms by which nanomaterials can cross lipid membranes. Apart from well-discussed energy-dependent endocytosis for large objects and passive diffusion through membranes by solute molecules, other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service