Skip to Content
Merck
All Photos(1)

Key Documents

700185M

Avanti

18:1 Chol (D7) ester

25,26,26,26,27,27,27-heptadeuteriocholest-5-en-3β-ol (9Z-octadecenoate), methylene chloride solution

Synonym(s):

111015

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H71D7O2
CAS Number:
Molecular Weight:
658.14
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (700185M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700185M

concentration

1 mg/mL (700185M-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1 Chol (D7) ester is a deuterated derivative of cholesteryl oleate (18:1 Cholester). 18:1 Chol (D7) ester is a component of SplashLipidoMix standard from Avanti and is used as an internal standard in ultrahigh performance liquid chromatography (UHPLC)-mass spectrometry (MS) analysis for profiling cerebrospinal fluid lipids.[1]

Biochem/physiol Actions

Cholesteryl oleate (18:1 Cholester) association with low-density lipoproteins (LDL) promotes atherosclerosis. The diet comprising monounsaturated fatty acids promotes cholesteryl ester secretion and association with lipoproteins.[2]

Packaging

5 mL Clear Glass Sealed Ampule (700185M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ane Iriondo et al.
Talanta, 195, 619-627 (2019-01-11)
The cerebrospinal fluid (CSF) lipidome is attracting increasing attention due to the importance of lipids in brain molecular signaling and their involvement in several neurological diseases. Different solvent systems have been used for the extraction of multiple lipid classes from
Chiara Degirolamo et al.
Journal of lipid research, 50 Suppl, S434-S439 (2008-11-26)
This review focuses on the relationships among dietary fat type, plasma and liver lipid, and lipoprotein metabolism and atherosclerosis. Dietary polyunsaturated fatty acids are beneficial for the prevention of coronary artery atherosclerosis. By contrast, dietary monounsaturated fatty acids appear to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service