Skip to Content
Merck
All Photos(1)

Key Documents

791647C

Avanti

15:0-18:1-d7 DG

Avanti Research - A Croda Brand 791647C

Synonym(s):

1-pentadecanoyl-2-oleyol(d7)-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H61D7O5
CAS Number:
Molecular Weight:
587.97
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791647C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791647C

concentration

1 mg/mL (791647C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

15:0-18:1-d7 DG (1-pentadecanoyl-2-oleyol(d7)-sn-glycerol) is a deuterated derivative of 15:0-18:1-DG, with deuteration in the oleic acid bonded to glycerol in sn-2 position. Pentadecanoic is bonded to sn-1 position. Diacylglycerols (DAG) are neutral lipids that carry fatty acid esterification. DAG is synthesized from the catabolic cleavage of headgroups of phospholipids. It is a small lipid and is membrane-associated.
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

15:0-18:1-d7 DG (1-pentadecanoyl-2-oleyol(d7)-sn-glycerol) has been used:
  • as an internal standard to spike skeletal muscle samples for liquid chromatography-mass spectroscopy (LC-MS) analysis
  • as a lipid standard in mass spectroscopy for the identification of lipids in mitochondria and lipid droplets
  • in the egg yolk lipid extraction as an internal standard for mass spectroscopy analysis

Biochem/physiol Actions

Diacylglycerols (DAG) are lipid messengers that participate in metabolic events. Altered levels, especially accumulation of DAG is implicated in insulin resistance. DAG serves as lipid modules for complex lipid synthesis. Protein kinase C (PKC) modulates DAG by interacting through its C1 domain. Impairment in DAG production is correlated to the pathology of Alzheimer′s disease, cancer and immune system diseases.

Packaging

2 mL Amber Glass Sealed Ampule (791647C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Silvia Carrasco et al.
Trends in biochemical sciences, 32(1), 27-36 (2006-12-13)
Diacylglycerol (DAG) has unique functions as a basic component of membranes, an intermediate in lipid metabolism and a key element in lipid-mediated signaling. In eukaryotes, for example, impaired DAG generation and/or consumption have severe effects on organ development and cell
Thomas Oliver Eichmann et al.
Cellular and molecular life sciences : CMLS, 72(20), 3931-3952 (2015-07-15)
The neutral lipids diacylglycerols (DAGs) are involved in a plethora of metabolic pathways. They function as components of cellular membranes, as building blocks for glycero(phospho)lipids, and as lipid second messengers. Considering their central role in multiple metabolic processes and signaling
Ya Xie et al.
Food chemistry, 319, 126547-126547 (2020-03-18)
In this research, a novel ultrasound-assisted extraction (USAE) method using a one-phase solvent (isopropanol, IPA) combined with liquid chromatography-electrospray ionization quadrupole time-of-flight mass spectrometry (LC-ESI-QToF-MS) was developed for exhaustive profiling of egg yolk lipids, which are considered to be essential
Paul T Reidy et al.
The Journal of physiology, 596(21), 5217-5236 (2018-09-09)
Insulin sensitivity (as determined by a hyperinsulinaemic-euglyceamic clamp) decreased 15% after reduced activity. Despite not fully returning to baseline physical activity levels, insulin sensitivity unexpectedly, rebounded above that recorded before 2 weeks of reduced physical activity by 14% after the recovery
Florian Geltinger et al.
GeroScience, 42(1), 19-38 (2019-11-05)
Originally Lipid droplets (LDs) were considered as being droplets for lipid storage only. Increasing evidence, however, demonstrates that LDs fulfill a pleiotropy of additional functions. Among them is the modulation of protein as well as lipid homeostasis. Under unfavorable pro-oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service