Skip to Content
Merck
All Photos(1)

Key Documents

860519P

Avanti

C18:1 Ceramide (d18:1/18:1(9Z))

Avanti Research - A Croda Brand 860519P, powder

Synonym(s):

N-oleoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H69NO3
CAS Number:
Molecular Weight:
563.94
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 25 mg (860519P-25mg)
pkg of 1 × 5 mg (860519P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860519P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C36H69NO3/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-36(40)37-34(33-38)35(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,29,31,34-35,38-39H,3-16,19-28,30,32-33H2,1-2H3,(H,37,40)/b18-17-,31-29+/t34-,35+/m0/s1

InChI key

OBFSLMQLPNKVRW-RHPAUOISSA-N

General description

C18:1 Ceramide (d18:1/18:1(9Z)) or N-oleoyl-D-erythro-sphingosine is a synthetic ceramide containing sphingosine with oleic acid.

Application

C18:1 Ceramide (d18:1/18:1(9Z)) may be used as a standard:
  • to quantify ceramide content in muscle tissue samples using high-performance liquid chromatography/tandem mass spectrometry (HPLC/MS2)[1]
  • for the absolute quantification of epidermal lipids using liquid chromatography-tandem mass spectrometry (LC/MS/MS)[2]
  • for the ratiometric quantification of sphingomyelin (SM)[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860519P-25mg)
5 mL Amber Glass Screw Cap Vial (860519P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meenu N Perera et al.
Biochimica et biophysica acta, 1818(5), 1291-1301 (2012-03-01)
The sphingolipid, ceramide, self-assembles in the mitochondrial outer membrane (MOM), forming large channels capable of translocating proteins. These channels are believed to be involved in protein release from mitochondria, a key decision-making step in cell death. Synthetic analogs of ceramide
Ayman J Aljohani et al.
Investigative ophthalmology & visual science, 55(6), 3413-3422 (2014-05-03)
We determined the profiles of sphingomyelin, sphingoid base, sphingoid base-1-phosphate, and ceramide, and their quantitative differences between control and glaucomatous trabecular meshwork (TM) derived from human donors. Control and primary open angle glaucoma (POAG) TM samples were collected from cadaver
Daniel A Peñalva et al.
Biochimica et biophysica acta, 1838(3), 731-738 (2013-12-10)
Unique species of ceramide (Cer) with very-long-chain polyunsaturated fatty acid (VLCPUFA), mainly 28-32 carbon atoms, 4-5 double bonds, in nonhydroxy and 2-hydroxy forms (n-V Cer and h-V Cer, respectively), are generated in rat spermatozoa from the corresponding sphingomyelins during the
Max Lönnfors et al.
Langmuir : the ACS journal of surfaces and colloids, 29(7), 2319-2329 (2013-01-30)
We prepared cholesteryl phosphocholine (CholPC) by chemical synthesis and studied its interactions with small (ceramide and cholesterol) and large headgroup (sphingomyelin (SM) and phosphatidylcholine) colipids in bilayer membranes. We established that CholPC could form bilayers (giant uni- and multilamellar vesicles
Donato A Rivas et al.
Journal of applied physiology (Bethesda, Md. : 1985), 113(11), 1727-1736 (2012-10-09)
One of the most fundamental adaptive physiological events is the response of skeletal muscle to high-intensity resistance exercise, resulting in increased protein synthesis and ultimately larger muscle mass. However, muscle growth in response to contraction is attenuated in older humans.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service