Skip to Content
Merck
All Photos(1)

Documents

W319309

Sigma-Aldrich

1-Methylnaphthalene

≥95%

Synonym(s):

α-methylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH3
CAS Number:
Molecular Weight:
142.20
FEMA Number:
3193
Beilstein:
506793
EC Number:
Council of Europe no.:
11009
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.014
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥95%

form

liquid

autoignition temp.

984 °F

refractive index

n20/D 1.615 (lit.)

bp

240-243 °C (lit.)

mp

−22 °C (lit.)

density

1.001 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

earthy

SMILES string

Cc1cccc2ccccc12

InChI

1S/C11H10/c1-9-5-4-7-10-6-2-3-8-11(9)10/h2-8H,1H3

InChI key

QPUYECUOLPXSFR-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

1-Methylnaphthalene occurs in Chinese raw spirits, brown millet, milled millet and millet bran. This volatile organic compound (VOC) is also found in the diesel engine exhaust gas and in the manures of cow, hog and chicken.

Application


  • Viscosities and Densities of Binary and Ternary Mixtures of Aliphatic and Polyaromatic Hydrocarbons: Pyrene +1-Methylnaphthalene + Dodecane at T = (293.15 to 343.15) K. Experiment and Modeling.: This research presents experimental data and modeling of the viscosities and densities of mixtures involving 1-methylnaphthalene, providing valuable data for industrial applications and theoretical studies in chemical engineering (Tenorio et al., 2024).

  • Chemical Composition and Optical Properties of Secondary Organic Aerosol from Photooxidation of Volatile Organic Compound Mixtures.: Investigates the secondary organic aerosols produced from photooxidation of volatile organic compounds including 1-methylnaphthalene, highlighting their optical properties and implications for air quality and climate modeling (Cui et al., 2024).

  • Oxidation 1-methyl naphthalene based on the synergy of environmentally persistent free radicals (EPFRs) and PAHs in particulate matter (PM) surface.: This study explores the oxidative mechanisms of 1-methyl naphthalene on particulate matter surfaces, facilitated by environmentally persistent free radicals. The findings contribute to the understanding of air pollution chemistry and environmental degradation processes (Ghimire et al., 2023).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile organic compounds and particulates as components of diesel engine exhaust gas.
Schulz H, et al.
Combustion and Flame, 118(1), 179-190 (1999)
Determination of the volatile composition in brown millet, milled millet and millet bran by gas chromatography/mass spectrometry.
Liu J, et al.
Molecules (Basel), 17(3), 2271-2282 (2012)
Identification of volatile organic compounds in the manures of cow, hog and chicken by solid phase microextraction coupled with gas chromatography/mass spectrometry
Huang J, et al.
Sepu, 25(3), 425-429 (2007)
Volatile compounds of raw spirits from different distilling stages of Luzhou-flavor spirit
Zheng, Jia, et al
Food Science and Technology Research, 20(2), 283-293 (2014)
Mykola Seredych et al.
Langmuir : the ACS journal of surfaces and colloids, 26(1), 227-233 (2009-12-30)
Two synthetic, polymer-derived carbons were modified with urea to incorporate nitrogen surface functional groups. Then they were investigated as adsorbents of dibenzothiophene (DBT) and 4, 6-dimethyldibenzothiophene (DMDBT) from simulated diesel fuel under dynamic conditions with the total concentration of sulfur

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service