Skip to Content
Merck
All Photos(2)

Key Documents

27500

Sigma-Aldrich

Triethyl citrate

greener alternative

≥98.0% (GC)

Synonym(s):

Ethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
Beilstein:
1801199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

Assay

≥98.0% (GC)

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

greener alternative category

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethyl citrate is a safe direct food additive[1].
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with "Less Hazardous Chemical Syntheses". Click here for more information.

Application

Triethyl citrate was used as a plasticizer in the study of the feasibility of near-infrared (NIR) spectroscopy in determining the influence of plasticizer levels and sintering temperature on the breaking force of extended-release matrix tablets prepared by roller-compaction[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monice M Fiume et al.
International journal of toxicology, 33(2 suppl), 16S-46S (2014-05-28)
The CIR Expert Panel (Panel) assessed the safety of citric acid, 12 inorganic citrate salts, and 20 alkyl citrate esters as used in cosmetics, concluding that these ingredients are safe in the present practices of use and concentration. Citric acid
Vivek S Dave et al.
Drug development and industrial pharmacy, 41(6), 898-905 (2014-05-03)
The aim of this study was to investigate the feasibility of near-infrared (NIR) spectroscopy for the determination of the influence of sintering temperature and plasticizer levels on the breaking force of extended-release matrix tablets prepared via roller-compaction. Six formulations using
Kakuyou Ogawa et al.
Biological & pharmaceutical bulletin, 37(9), 1559-1563 (2014-09-02)
Zerumbone 1 is an 11-membered cyclic sesquiterpene obtained from the rhizomes of Zingiber zerumbet SMITH (Zingiberaceae). In this study, we investigated the structure-activity relationship of 1, α-humulene (2), tetrahydrozerumbone stereoisomers (3-5), and tetrahydrozerumbone derivatives (6-9). The oxygen-containing functional groups and
S Monfort et al.
International journal of food microbiology, 145(2-3), 476-482 (2011-02-19)
This paper is an evaluation of the lethal effectiveness of a successive application of pulsed electric fields (PEFs) and heat treatment in liquid whole egg (LWE) in the presence of different additives on the population of Salmonella serovar Enteritidis. Synergistic
Gavin P Andrews et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 69(1), 264-273 (2008-01-01)
The aim of this highly novel study was to use hot-melt extrusion technology as an alternative process to enteric coating. In so doing, oral dosage forms displaying enteric properties may be produced in a continuous, rapid process, providing significant advantages

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service