Skip to Content
Merck
All Photos(1)

Documents

E40005

Sigma-Aldrich

2-Ethylnaphthalene

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7C2H5
CAS Number:
Molecular Weight:
156.22
Beilstein:
1903545
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.599 (lit.)

bp

251-252 °C (lit.)

mp

−70 °C (lit.)

density

0.992 g/mL at 25 °C (lit.)

fluorescence

λex 300 nm; λem 335 nm in ethanol

SMILES string

CCc1ccc2ccccc2c1

InChI

1S/C12H12/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h3-9H,2H2,1H3

InChI key

RJTJVVYSTUQWNI-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Joong Kang et al.
Chemosphere, 222, 656-664 (2019-02-08)
Crude oil released into the environment contains many polycyclic aromatic hydrocarbons (PAHs). Alkylated PAHs are more abundant than unsubstituted PAHs and their toxicity is also of serious concern. Among the various physical, chemical, and biological weathering processes of crude oils
J M Shoulder et al.
SAR and QSAR in environmental research, 30(4), 229-245 (2019-03-22)
Persistent organic contaminants in the environment pose an environmental risk due to widespread occurrence and toxic properties. Advanced oxidation processes (AOPs) are treatment methods that have been used to successfully degrade organic contaminants in water, soil, sediments and sludge. Reaction
Pengfei Liu et al.
Polymers, 11(2) (2019-04-10)
Covalently crosslinked nanogels are widely explored as drug delivery systems and sensors. Radical polymerization provides a simple, inexpensive, and broadly applicable approach for their preparation, although the random nature of the reaction requires careful study of the final chemical composition.
Manickam Selvaraj et al.
Dalton transactions (Cambridge, England : 2003), 49(28), 9710-9718 (2020-07-03)
A very green catalytic method has been introduced for the synthesis of alkylaromatic ketones by solvent-free benzylic oxidation of alkylaromatics with molecular oxygen (O2) over hexagonally mesostructured MnSBA-15 catalysts synthesized with a variety of manganese (Mn) contents using a pH-adjusting
Danlei Wang et al.
Chemico-biological interactions, 315, 108905-108905 (2019-11-26)
Mineral oils are widely applied in food production and processing and may contain polycyclic aromatic hydrocarbons (PAHs). The PAHs that may be present in mineral oils are typically alkylated, and have been barely studied. Metabolic oxidation of the aromatic ring

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service