Skip to Content
Merck
All Photos(1)

Key Documents

W254711

Sigma-Aldrich

Heptyl acetate

natural, ≥98%, FG

Synonym(s):

n-Heptyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2547
EC Number:
Council of Europe no.:
212c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.022
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5.5 (vs air)

vapor pressure

12 mmHg ( 73 °C)

Assay

≥98%

refractive index

n20/D 1.415 (lit.)

bp

192-193 °C (lit.)

mp

−50.2 (lit.)

density

0.886 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; waxy; citrus; woody; wine-like

SMILES string

CCCCCCCOC(C)=O

InChI

1S/C9H18O2/c1-3-4-5-6-7-8-11-9(2)10/h3-8H2,1-2H3

InChI key

ZCZSIDMEHXZRLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Isitt et al.
Journal of chemical ecology, 46(5-6), 497-507 (2020-06-20)
Prior research from trapping experiments using synthetic pheromone components suggested the existence of local and geographical variation in the pheromone system of the spruce beetle, Dendroctonus rufipennis (Kirby). To test hypotheses concerning variation in the spruce beetle's aggregation pheromone blend
Letian Xu et al.
Microbial ecology, 70(4), 1012-1023 (2015-05-20)
Bark beetles are the most important mortality agent in coniferous forests, and pheromones play important roles in their management. Dendroctonus valens LeConte was introduced from North America to China and has killed millions of healthy pines there. Trapping with semiochemicals
Jenny M Lindh et al.
Malaria journal, 14, 119-119 (2015-04-18)
New strategies are needed to manage malaria vector populations that resist insecticides and bite outdoors. This study describes a breakthrough in developing 'attract and kill' strategies targeting gravid females by identifying and evaluating an oviposition attractant for Anopheles gambiae s.l.
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service