Merck
Mosquito odorant receptor for DEET and methyl jasmonate.
Proceedings of the National Academy of Sciences of the United States of America (2014-10-29)
Pingxi Xu, Young-Moo Choo, Alyssa De La Rosa, Walter S Leal
ABSTRACT
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes use their sense of smell to detect DEET, but there are currently two hypotheses regarding its mode of action: activation of ionotropic receptor IR40a vs. odorant receptor(s). Here, we demonstrate that DEET, picaridin, insect repellent 3535, and p-menthan-3,8-diol activate the odorant receptor CquiOR136 of the southern house mosquito, Culex quinquefasciatus. Electrophysiological and behavioral assays showed that CquiIR40a knockdown had no significant effect on DEET detection and repellency. By contrast, reduction of CquiOR136 transcript levels led to a significant decrease in electroantennographic responses to DEET and a complete lack of repellency. Thus, direct activation of an odorant receptor, not an ionotropic receptor, is necessary for DEET reception and repellency in Culex mosquitoes. Interestingly, methyl jasmonate, a repellent derived from the nonvolatile jasmonic acid in the signaling pathway of plant defenses, elicited robust responses in CquiOR136•CquiOrco-expressing Xenopus oocytes, thus suggesting a possible link between natural products with long insect-plant evolutionary history and synthetic repellents.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Geranyl acetate, ≥97%
Sigma-Aldrich
2,3-Dimethylphenol, 98%
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
2-Undecanone, 99%
Supelco
Isobutyric acid, analytical standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Nonanal, analytical standard
Sigma-Aldrich
Lauric aldehyde, natural, ≥95%, FG
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Sigma-Aldrich
cis-3-Hexenyl acetate, ≥98%, stabilized, FCC, FG
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Supelco
Melting point standard 79-81°C, analytical standard
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Tiglic aldehyde, ≥96%
Sigma-Aldrich
Isoamyl acetate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Heptanoic acid, ≥99.0% (GC)
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology