Skip to Content
Merck
All Photos(3)

Key Documents

W241415

Sigma-Aldrich

Ethyl acetate

greener alternative

natural, ≥99%, FCC, FG

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2414
Beilstein:
506104
EC Number:
Council of Europe no.:
191
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.001
NACRES:
NA.21
grade:
FG
Kosher
natural
food allergen:
no known allergens

grade

FG
Kosher
natural

Quality Level

300
400

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

801 °F

shelf life

60 mo.

expl. lim.

2.2-11.5 %, 38 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

format

neat

greener alternative category

Organoleptic

fruity; ethereal

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate can be used as a flavoring agent in the food industry. It is one of the volatile components identified in Spirulina platensis.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • New piperazine derivatives helvamides B-C from the marine-derived fungus Penicillium velutinum ZK-14 uncovered by OSMAC (One Strain Many Compounds) strategy.: Ethyl acetate is utilized as a solvent to isolate new bioactive compounds from marine fungi, indicating its crucial role in drug discovery and natural product research (Borkunov et al., 2024).

  • Thermodynamic Analysis of Eplerenone in 13 Pure Solvents at Temperatures from 283.15 to 323.15 K.: This article provides comprehensive data on the solubility and thermodynamics of pharmaceutical compounds in ethyl acetate, demonstrating its importance in pharmaceutical formulations (Liu et al., 2024).

  • Disruption of NADPH Homeostasis by Total Flavonoids from Adinandra nitida Merr. ex Li Leaves Triggers ROS-Dependent p53 Activation Leading to Apoptosis in Non-Small Cell Lung Cancer Cells.: Ethyl acetate is used to extract bioactive flavonoids, contributing to research in cancer treatment by demonstrating the pro-apoptotic effects on lung cancer cells (Lan et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial activity of volatile component and various extracts of Spirulina platensis
Ozdemir G, et al.
Phytotherapy Research, 18(9), 754-757 (2004)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 940-941 (1997)
Junwei Liu et al.
Planta, 241(6), 1435-1451 (2015-02-27)
Strigolactone changes and cross talk with ABA unveil a picture of root-specific hormonal dynamics under stress. Strigolactones (SLs) are carotenoid-derived hormones influencing diverse aspects of development and communication with (micro)organisms, and proposed as mediators of environmental stimuli in resource allocation
K J S Anand et al.
British journal of anaesthesia, 101(5), 680-689 (2008-08-30)
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100
Yuusuke Fujimoto et al.
PloS one, 7(11), e48685-e48685 (2012-11-13)
Combination therapy with ribavirin, interferon, and viral protease inhibitors could be expected to elicit a high level of sustained virologic response in patients infected with hepatitis C virus (HCV). However, several severe side effects of this combination therapy have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service