STW6
COware gas reactor
reaction scale of 5.0-20.0 mmol (total volume 400 mL)
About This Item
Recommended Products
packaging
pack of 1 ea
General description
- 0.1-1 mmol (total volume 20 mL)
- 1.0-5.0 mmol (total volume 100 mL)
- 5.0-20.0 mmol (total volume 400 mL)
Each unit includes two screw caps, two stabilizing PTFE discs (recyclable), ten single use silicone/PTFE septa (in combination with the stabilizing discs each septum allows for a maximum of 2 piercings using 21G/Ø0.80 mm syringes or smaller).
Order replacement PTFE/silicone septa STW3.
Learn more about CO releasing molecules from Prof. Skrydstrup
COware gas reactor is a part of the COware Platform
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Articles
Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.
Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.
Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.
Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.