Skip to Content
Merck
All Photos(2)

Key Documents

D210803

Sigma-Aldrich

Diphenyl phosphite

Synonym(s):

Diphenoxyphosphine oxide, Diphenyl phosphonate, NSC 43786

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2P(O)H
CAS Number:
Molecular Weight:
234.19
Beilstein:
2051909
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.1 (vs air)

Quality Level

vapor pressure

5 mmHg ( 140 °C)

form

liquid

impurities

≤15% phenol

refractive index

n20/D 1.558 (lit.)

bp

218-219 °C/26 mmHg (lit.)

mp

12 °C (lit.)

density

1.223 g/mL at 25 °C (lit.)

SMILES string

O=[PH](Oc1ccccc1)Oc2ccccc2

InChI

1S/C12H11O3P/c13-16(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10,16H

InChI key

OGBPILLJZSJJRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of N-benzylarylamide and di-Ph α-aminomethanephosphonate derivatives used as additives for thermooxidative stability of ester-based oils
  • One-pot three-component Kabachnik-Fields reactions
  • Visible light mediated oxidative phosphonylations of amines
  • Modified asymmetric Pudovik addition with haloketones
  • Tandem decarboxylation-coupling with natural alpha-amino acids

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenhua Fang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 85, 160-168 (2016-12-09)
Osteosarcoma is the most common primary malignant bone tumor in the pediatric age group, and chemotherapy directed by targeted nanoparticulate drug delivery system represents a promising approach for osteosarcoma treatment recently. Here, we designed and developed a novel DOX-loaded targeted
Jiesheng Xv et al.
Journal of biomaterials science. Polymer edition, 30(5), 398-414 (2019-01-29)
Zwitterionic polymer is a new generation of anti-fouling materials with its good resistance to protein and bacterial adhesion. Constructing the anti-fouling surfaces with zwitterionic polymer has been regarded as an effective approach for improving the biocompatibility and biofunctionality of clinic
Diana Rogacz et al.
Materials (Basel, Switzerland), 12(12) (2019-06-27)
The aim of this work was to evaluate the impact of the thiophene-derived aminophosphonates 1-6 on seedling emergence and growth of monocotyledonous oat (Avena sativa) and dicotyledonous radish (Raphanus sativus L.), and phytotoxicity against three persistent and resistant weeds (Galinsoga
Diana Rogacz et al.
Molecules (Basel, Switzerland), 23(12) (2018-12-06)
Background: The aim of this work was to evaluate phytotoxicity of the thiophene derivatives against three persistent weeds of a high degree of resistance (Galinsogaparviflora Cav.,Rumexacetosa L., and Chenopodiumalbum) as well as their ecotoxicological impact on Heterocypris incongruens. In addition
Ye He et al.
International journal of biological macromolecules, 163, 1738-1746 (2020-09-18)
A zwitterionic phosphoryldiserine (PDS)- chitosan conjugate was synthesized via Atherton-Todd reaction, and its degree of substitution of PDS and structure were characterized by 1H and 31P NMR spectra, ICP, FTIR and XRD. Thermal analysis confirmed that there existed the freezing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service