Skip to Content
Merck
All Photos(2)

Key Documents

D178454

Sigma-Aldrich

Dimethyl phosphite

98%

Synonym(s):

Dimethoxyphosphine oxide, Dimethyl hydrogen phosphite, Dimethyl hydrogen phosphonate, Dimethyl phosphonate, Hydrogen dimethyl phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)H
CAS Number:
Molecular Weight:
110.05
Beilstein:
1697490
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

impurities

≤0.5% trimethyl phosphate
≤1% methyl alcohol

refractive index

n20/D 1.402 (lit.)

bp

170-171 °C (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

[H]P(=O)(OC)OC

InChI

1S/C2H7O3P/c1-4-6(3)5-2/h6H,1-2H3

InChI key

HZCDANOFLILNSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl phosphite is used in the synthesis of α-aminophosphonates, which are medicinally important phosphorus analogs of amino acids, by treating with an amine and an aldehyde or a ketone. It can also be employed in asymmetric hydrophosphonylation of aldehydes to synthesize optically active α-hydroxy phosphonates.
Dimethyl phosphite may be used as an effective alternative to organic tin hydrides as radical reducing agents for organic halides, thionoesters, and isocyanides to form the corresponding hydrocarbons. Radical translocation/cyclization reactions of terminal alkynes to form cyclic phosphonates can also be effectively carried using dimethyl phosphite as a reagent under tin-free condition.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted solvent-free and catalyst-free Kabachnik?Fields reactions for α-amino phosphonates.
Mu X J, et al.
Tetrahedron Letters, 47(7), 1125-1127 (2006)
Dimethyl Phosphite Mediated Hydrogen Atom Abstraction: A Tin?Free Procedure for the Preparation of Cyclopentane Derivatives.
Beaufils F, et al.
Angewandte Chemie (International Edition in English), 44(33), 5273-5275 (2005)
Synthesis of an Optically Active C1?Symmetric Al (salalen) Complex and Its Application to the Catalytic Hydrophosphonylation of Aldehydes.
Saito B and Katsuki T
Angewandte Chemie (International Edition in English), 117(29), 4676-4678 (2005)
Zirconium (IV) compounds as efficient catalysts for synthesis of α-aminophosphonates.
Bhagat S and Chakraborti A K
The Journal of Organic Chemistry, 73(15), 6029-6032 (2008)
Ming Xie et al.
Journal of hazardous materials, 181(1-3), 975-980 (2010-06-18)
Nanofiltration (NF) was investigated for the removal of glyphosate in the neutralization liquor produced by the glycine-dimethylphosphit process. The Desal-5 DK membrane was chosen as the most suitable membrane for the NF process when compared to the DL and NTR7450

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service