Skip to Content
Merck
All Photos(1)

Key Documents

D102504

Sigma-Aldrich

1,5-Difluoro-2,4-dinitrobenzene

97%

Synonym(s):

DFDNB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H2(NO2)2
CAS Number:
Molecular Weight:
204.09
Beilstein:
1883116
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

72-74 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(c(F)cc1F)[N+]([O-])=O

InChI

1S/C6H2F2N2O4/c7-3-1-4(8)6(10(13)14)2-5(3)9(11)12/h1-2H

InChI key

VILFTWLXLYIEMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yanshun Liu et al.
Protein science : a publication of the Protein Society, 11(2), 371-380 (2002-01-16)
When concentrated in mildly acidic solutions, bovine pancreatic ribonuclease (RNase A) forms long-lived oligomers including two types of dimer, two types of trimer, and higher oligomers. In previous crystallographic work, we found that the major dimeric component forms by a
Alexander Shivanyuk et al.
Organic letters, 4(9), 1555-1558 (2002-04-27)
[reaction: see text]. o-alkylation of C2V-symmetrical resorcinarene tetraesters 2 with 2 equiv of 1,3-difluoro-4,6-dinitrobenzene readily affords conformationally rigid octanitro resorcinarene 3, which is a potential scaffold for the design of supramolecular structures.
S Simonsson et al.
The Journal of biological chemistry, 273(38), 24633-24639 (1998-09-12)
The UL9 gene of herpes simplex virus type 1 (HSV-1) encodes an origin binding protein (OBP). It is an ATP-dependent DNA helicase and a sequence-specific DNA-binding protein. The latter function is carried out by the C-terminal domain of OBP (DeltaOBP).
Zhanguo Wang et al.
Journal of combinatorial chemistry, 9(4), 652-660 (2007-05-17)
This paper discusses the synthesis of privileged structures 4H-benzo[1,4]thiazin-3-one and 1,1-dioxo-1,4-dihydro-2H-1lambda6-benzo[1,4]thiazin-3-one derivatives in a parallel solution-phase manner using 1,5-difluoro-2,4-dinitrobenzene. Each scaffold possesses four diversity points. A cheap and efficient oxidant, urea-hydrogen peroxide (UHP), was applied for the introduction of the
Yunyun Yuan et al.
Journal of combinatorial chemistry, 9(1), 158-170 (2007-01-09)
This paper discusses the synthesis of benzo[1,4]oxazin-3-one-based compounds from 1,5-difluoro-2,4-dinitrobenzene (1), including benzo[1,4]oxazin-3-ones (5-11) and five novel benzo[1,4]oxazin-3-one-based tricycles: 6-hydroxy-4H-1-oxa-4,5,8-triazaanthracen-3-one (14), 3,8-dihydro-5-oxa-1,3,8-triazacyclopenta-[b]-naphthalene-7-one (15, 17, 21), 3,8-dihydro-5-oxa-1,2,3,8-tetraazacylopenta[b]-naphthalene-7-one (16, 20), 3,8-dihydro-1H-5-oxa-1,3,8-triazacyclopenta[b]-naphthalene-2,7-dione (18, 22), and 5,8-dihydro-4H-1-oxa-4,5,8-triazaanthracene-3,6,7-trione (19). Finally, a chemical library based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service