Skip to Content
Merck
All Photos(2)

Key Documents

C112909

Sigma-Aldrich

2-Cyclopenten-1-one

98%

Synonym(s):

1-Cyclopenten-3-one, 1-Cyclopenten-5-one, 2-Cyclopentenone, Cyclopent-2-en-1-one, Cyclopenten-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
Beilstein:
1446054
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

64-65 °C/19 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CCC=C1

InChI

1S/C5H6O/c6-5-3-1-2-4-5/h1,3H,2,4H2

InChI key

BZKFMUIJRXWWQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile electrophile employed in a variety of addition reactions including conjugate addition of organocopper nucleophiles,[1] Michael reaction with silyl enol ethers,[2] and siloxanes,[3] Diels-Alder cycloadditions,[4] and phosphoniosilylations.[5]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brett D Schwartz et al.
Organic letters, 15(8), 1934-1937 (2013-04-05)
The title natural product, 1, has been synthesized in 20 steps from the enantiomerically pure cis-1,2-dihydrocatechol 2, itself obtained through the whole-cell biotransformation of toluene. The pivotal steps in the reaction sequence involve a Diels-Alder cycloaddition reaction between diene 2
Rachel Lerebours et al.
Organic letters, 9(14), 2737-2740 (2007-06-15)
Palladium-phosphinous acids catalyze the conjugate addition of arylsiloxanes to a wide range of alpha,beta-unsaturated substrates in water. A microwave-assisted procedure is described that uses 5 mol % of POPd1 to afford beta-substituted ketones, aldehydes, esters, nitriles, and nitroalkanes in 83%
Diverse reactivity in a rhodium(III)-catalyzed oxidative coupling of N-allyl arenesulfonamides with alkynes.
Dongqi Wang et al.
Angewandte Chemie (International ed. in English), 51(49), 12348-12352 (2012-10-31)
Xiaoxun Li et al.
Organic letters, 14(6), 1584-1587 (2012-03-03)
Functionalized cyclopentenones were synthesized by a Rh-catalyzed carbonylation of 3-acyloxy-1,4-enynes, derived from alkynes and α,β-unsaturated aldehydes. The reaction involved a Saucy-Marbet 1,3-acyloxy migration of propargyl esters and a [4 + 1] cycloaddition of the resulting acyloxy substituted vinylallene with CO.
Daniel J Kerr et al.
Organic letters, 14(7), 1732-1735 (2012-03-30)
Oxazolidinones are powerful promoters of the Nazarov reaction, enabling the cyclization of conventionally resistant substrates to be achieved under mild conditions. They exert excellent regio- and torquoselective control in both the conventional Nazarov reaction giving cyclopentenones and in the "interrupted"

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service