Skip to Content
Merck
All Photos(1)

Key Documents

P55708

Sigma-Aldrich

(R)-(+)-Pulegone

85%, technical grade

Synonym(s):

(+)-Pulegone, (R)-2-Isopropylidene-5-methylcyclohexanone, (R)-p-Menth-4(8)-en-3-one, p-Menth-4(8)-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2040703
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor pressure

138 mmHg ( 25 °C)

Assay

85%

optical activity

[α]20/D +22°, neat

refractive index

n20/D 1.486 (lit.)

bp

224 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CC\C(=C(\C)C)C(=O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-Pulegone, a monoterpene ketone found in essential oil of pennyroyal, can induce abortion.

Application

(R)-(+)-Pulegone may be used as a starting material to synthesize:
  • (2S,4R,6R,8S)-2,4,8-trimethyl-1,7-dioxaspiro[5.5]undecane, a spiroketal
  • (R)-(+)-citronellic acid, an intermediate to prepare leucine-d3
  • (+)-fawcettidine, a Lycopodium alkaloid

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From (R)-(+)-pulegone to (2S,4R,6R,8S)-2,4,8-trimethyl-1,7-dioxaspiro[5.5]undecane-A unique spiroacetal from the insect Kingdom.
Tu YQ, et al.
Tetrahedron Asymmetry, 6(2), 397-400 (1995)
Total Synthesis of (+)-Fawcettidine.
Kozak JA and Dake GR.
Angewandte Chemie (International Edition in English), 47(22), 4221-4223 (2008)
Metabolism of (R)-(+)-pulegone in F344 rats.
Chen LJ, et al.
Drug Metabolism and Disposition, 29(12), 1567-1577 (2001)
Synthesis of (2S, 4S)-and (2S, 4R)-[5,5,5-2H3] leucine from (R)-pulegone.
Hill RK, et al.
Canadian Journal of Chemistry, 72(1), 110-113 (1994)
Maung Kyaw Moe Tun et al.
The Journal of organic chemistry, 77(20), 9422-9425 (2012-10-04)
A three-step synthesis of (R)-(+)-4-methylcyclohex-2-ene-1-one (1) from (R)-(+)-pulegone (3), proceeding in 44% overall yield, is described. The sequence comprises vinyl triflate formation, site-selective ozonolysis, and reduction. The route requires only one chromatographic purification and provides a convenient method to access

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service