Skip to Content
Merck
All Photos(1)

Key Documents

160687

Sigma-Aldrich

2-Cyclohepten-1-one

80%, technical grade

Synonym(s):

2-Cycloheptenone, Tropilene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H10(=O)
CAS Number:
Molecular Weight:
110.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

80%

form

liquid

refractive index

n20/D 1.494 (lit.)

density

0.988 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

O=C1CCCCC=C1

InChI

1S/C7H10O/c8-7-5-3-1-2-4-6-7/h3,5H,1-2,4,6H2

InChI key

WZCRDVTWUYLPTR-UHFFFAOYSA-N

General description

2-Cyclohepten-1-one is an α,β-enone and its regioselective reaction with allyl indium reagent in the presence of TMSCl has been reported. It reacts with Bu2Zn in the presence of catalytic amounts of Cu(OTf)2 and CH2-bridged azolium salts to give (S)-3-butylcycloheptanone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Masukawa et al.
Life sciences, 44(6), 417-424 (1989-01-01)
To search for a technique to deplete reduced glutathione (GSH) in brain, the influence of various types of compounds on brain GSH levels was investigated in mice. Of the compounds tested, cyclohexene-1-one, cycloheptene-1-one and diethyl maleate were shown to be
G Benzi et al.
Neurobiology of aging, 12(3), 227-231 (1991-05-01)
A severe age-dependent depletion of reduced glutathione (GSH) occurs in rat forebrain at 1-3 h from intraperitoneal injection of the electrophilic agents cyclohexene-1-one and cycloheptene-1-one. Chronic pretreatment with central dopamine agonists (i.e., ergot alkaloids; particularly, dihydroergocriptine) partially counteracts the GSH
Naoatsu Shibata et al.
The Journal of organic chemistry, 77(8), 4079-4086 (2012-03-30)
A series of hydroxy-amide functionalized azolium salts have been designed and synthesized for Cu-catalyzed asymmetric conjugate addition reaction. The (CH(2))(2)-bridged hydroxy-amide functionalized azolium ligand precursors 2, in addition to the previously reported CH(2)-bridged azolium salts 1, have been prepared from
P R Byron et al.
Journal of pharmaceutical sciences, 69(5), 527-531 (1980-05-01)
A stirred transfer cell containing equal volumes of light liquid paraffin and an aqueous phase at 37 degrees was used to demonstrate the feasibility of calculating the partition coefficient of an unstable compound by kinetic analysis. Cyclohept-2-enone was chosen since
Kleber Thiago de Oliveira et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 63(3), 709-713 (2005-07-19)
A detailed analysis with total assignment of (1)H and (13)C NMR spectral data for a cycloheptenone derivative, a key intermediate for the synthesis of perhydroazulene terpenoids, is related. These assignments are based on 1D (1)H and (13)C NMR and on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service