Skip to Content
Merck
All Photos(3)

Documents

B89205

Sigma-Aldrich

tert-Butylamine

98%

Synonym(s):

TBA, 2-Amino-2-methylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
605267
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.5 (vs air)

Quality Level

100
300

vapor pressure

5.7 psi ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

716 °F

expl. lim.

9.8 %

refractive index

n20/D 1.377 (lit.)

bp

46 °C (lit.)

mp

−67 °C (lit.)

density

0.696 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)N

InChI

1S/C4H11N/c1-4(2,3)5/h5H2,1-3H3

InChI key

YBRBMKDOPFTVDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butylamine is an organic chemical compound used as an intermediate in the synthesis of sulfenamides.

Application

tert-Butylamine (TBA) can be used in the following applications:
  • It is an effective solvent for the removal of surfactants such as PVP and capping agents such as KBr from the nanoparticle surface.
  • Mixed complexes of platinum, amino acid and TBA show potent anti-tumor activities.
  • It is an effective base for the aziridination of 2-arylethenyl(diphenyl)sulfonium salt to form the corresponding N-tert-butylaziridine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-36.4 °F - closed cup

Flash Point(C)

-38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antitumor complexes of platinum with carrier molecules. 3. Cytotoxicity of some platinum amino acid complexes against cisplatin-sensitive and resistant L1210 leukemia cells.
Colombo A, et al.
Inorgorganica Chimica Acta, 125(1), L1-L3 (1986)
Removal of surfactant and capping agent from Pd nanocubes (Pd-NCs) using tert-butylamine: its effect on electrochemical characteristics.
Naresh N, et al.
Journal of Material Chemistry A, 1(30), 8553-8559 (2013)
Copper-Catalyzed Formation of Sulfur?Nitrogen Bonds by Dehydrocoupling of Thiols with Amines
Taniguchi N
European Journal of Organic Chemistry (2010)
Antitumor complexes of platinum with carrier molecules. 2 [1]. Mixed complexes of amino acids and tert-butylamine.
Bersanetti E, et al.
Inorgorganica Chimica Acta, 93(4), 167-172 (1984)
A convenient method for the synthesis of 2-arylaziridines from styrene derivatives via 2-arylethenyl (diphenyl) sulfonium salts.
Matsuo J I, et al.
Chemistry Letters (Jpn), 32(4), 392-393 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service