Skip to Content
Merck
All Photos(1)

Key Documents

BI0832

Sigma-Aldrich

Deblocking Reagent 3% Dichloroacetic Acid in Toluene

Novabiochem®

Synonym(s):

DCA Deblock, dna

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
MDL number:
UNSPSC Code:
41116006
Pricing and availability is not currently available.

grade

reagent

Quality Level

200
300

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: Lewis acid

color

APHA: ≤20

transition temp

flash point 4 °C

shipped in

ambient

storage temp.

2-30°C

SMILES string

OC(=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BI0832-1005 and BI0832-2505 are packaged in GL45 glass bottles
BI0832-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Novabiochem®

Analysis Note

Color (APHA)20 maxComponent: Toluene Purity99.5% minDensity at 20°Cg/mlDichloroacetic acid2.95-3.05% v/vFormClear liquidWater (H2O)300ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service